Skip to Content
Merck
All Photos(1)

Key Documents

UC18

Sigma-Aldrich

(±)-Geosmin

≥97% (GC), liquid (oil)

Synonym(s):

2β,6α-Dimethylbicyclo[4.4.0]decan-1β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O
CAS Number:
Molecular Weight:
182.30
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

product name

(±)-Geosmin, ≥97% (GC)

Assay

≥97% (GC)

form

liquid (oil)

color

colorless

storage temp.

2-8°C

SMILES string

CC1CCCC2(C)CCCCC12O

InChI

1S/C12H22O/c1-10-6-5-8-11(2)7-3-4-9-12(10,11)13/h10,13H,3-9H2,1-2H3

InChI key

JLPUXFOGCDVKGO-UHFFFAOYSA-N

Application

(±)-Geosmin has been used as a standard for GC-MS based analytical techniques. Geosmin has also been used to study its adsorption on activated carbon.

Biochem/physiol Actions

(±)-Geosmin is produced by Streptomyces coelicolor and cyanobacteria.
Geosmin is a naturally occurring biomolecule that contributes to the earthy or musty flavor and odor in water supplies. This water contaminant can be detected by humans at ng/l (parts per trillion) concentrations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nayra Rodrigues de Alcântara et al.
Applied microbiology and biotechnology, 104(11), 5065-5080 (2020-04-08)
Mycobacterium abscessus subsp. massiliense (Mycma) belongs to the Mycobacterium abscessus complex and is a rapidly growing non-tuberculous mycobacterium. The chronic pulmonary, skin, and soft tissue infections that it causes may be difficult to treat due to its intrinsic resistance to
O Antonoglou et al.
Materials science & engineering. C, Materials for biological applications, 99, 264-274 (2019-03-21)
Heterometal oxide nanoparticles of bioessential metals are shedding new light to nanoparticle-inspired bioapplications. Pairing bioreactive elements like copper and iron can affect the redox dynamic and biological profile of the nanomaterial. Given the complexity of physicochemical properties, biological activity and
Ezaz Ahmad et al.
Biochemical and biophysical research communications, 522(1), 226-232 (2019-11-25)
In Escherichia coli, G/C-rich hairpin structure followed by a U-tract in the 3' region of the nascent RNA are crucial determinants for intrinsic or factor independent transcription termination. In mycobacteria, there is a scarcity of such intrinsic terminators. However, secondary
Flávio Henrique Jesus-Santos et al.
Frontiers in cellular and infection microbiology, 10, 408-408 (2020-09-10)
On the surface of the Leishmania promastigote, phosphoglycans (PG) such as lipophosphoglycan (LPG), proteophosphoglycan (PPG), free phosphoglycan polymers (PGs), and acid phosphatases (sAP), are dominant and contribute to the invasion and survival of Leishmania within the host cell by modulating
S W Lloyd et al.
Journal of agricultural and food chemistry, 47(1), 164-169 (1999-11-24)
The semivolatile cyclic alcohols 2-methylisoborneol (MIB) and geosmin (GSM) impart muddy or musty flavors to water and food products. A rapid quantitative analytical technique has been developed whereby microwave distillation is used to remove the volatile organic compounds from a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service