Skip to Content
Merck
All Photos(3)

Key Documents

D70201

Sigma-Aldrich

2,6-Dichlorophenol

99%

Synonym(s):

2,6-Dichlorophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein:
1447806
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

crystals

bp

218-220 °C (lit.)

mp

64-66 °C (lit.)

SMILES string

Oc1c(Cl)cccc1Cl

InChI

1S/C6H4Cl2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H

InChI key

HOLHYSJJBXSLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Tian et al.
Water research, 47(1), 279-288 (2012-11-06)
Distribution variation of a metabolic uncoupler, 2,6-dichlorophenol (2,6-DCP), in long-term sludge culture was studied, and the effects on sludge reduction and biological inhibition of this chemical during the 90-day operation were established. The extracellular polymeric substance (EPS) matrix functioned as
Anna Maria Polcaro et al.
Journal of hazardous materials, 148(3), 505-512 (2007-04-07)
This paper presents the results of a study on the electrokinetic treatment of kaolinite and humic acid kaolinite complexes spiked with 2,6-dichlorophenol or 3-(3,4-dichlorophenyl)-1,1-dimethylurea (diuron). In particular, the attention was paid to the interaction between solid surface and dissolved organics:
S Almeda et al.
Analytica chimica acta, 587(1), 97-103 (2007-03-28)
Actually there is a great trend on the development of effective analytical methods for monitoring trace levels of various phenols which can indicate, among others compounds, the water quality. A simple, inexpensive supported liquid membrane (SLM) device was used in
Lígia Miranda Ferreira Borges et al.
Veterinary parasitology, 147(1-2), 155-160 (2007-04-27)
This study was carried out with the objective of evaluating the efficacy of a 2,6-dichlorophenol (2,6-DCP) lure to control Dermacentor nitens (Acari: Ixodidae). Slow-release formulations of the pheromone formulated with and without cypermethrin were prepared. Olfactometer bioassays were used to
Davide Vione et al.
Chemosphere, 69(10), 1548-1554 (2007-07-10)
2,4-Dichlorophenol (2,4-DCP) and 2,6-dichlorophenol (2,6-DCP) undergo oxidation, nitrosation and nitration in the presence of nitrate under UV irradiation. Nitration is favoured under acidic conditions, most likely because HNO(2) is formed on nitrate photolysis. The most likely photonitration pathway is the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service