Skip to Content
Merck
All Photos(1)

Documents

UC166

Sigma-Aldrich

Sulfaphenazole

≥98% (HPLC)

Synonym(s):

4-Amino-N-(1-phenyl-1H-pyrazol-5-yl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N4O2S
CAS Number:
Molecular Weight:
314.36
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white to light yellow

mp

179-183 °C

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2ccnn2-c3ccccc3

InChI

1S/C15H14N4O2S/c16-12-6-8-14(9-7-12)22(20,21)18-15-10-11-17-19(15)13-4-2-1-3-5-13/h1-11,18H,16H2

InChI key

QWCJHSGMANYXCW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Antibacterial. Specific inhibitor of CYP2C9. Blocks pro-inflammatory and atherogenic effects of linoleic acid (increase in oxidative stress and activation of AP-1) mediated by CYP2C9. Specific inhibitor of CYP2C9. Blocks pro-inflammatory and atherogenic effects of linoleic acid (increase in oxidative stress and activation of AP-1) mediated by CYP2C9. Inhibits bradykinin-induced tPA release.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chengqun Huang et al.
American journal of physiology. Heart and circulatory physiology, 298(2), H570-H579 (2009-12-17)
Previously, we showed that sulfaphenazole (SUL), an antimicrobial agent that is a potent inhibitor of cytochrome P4502C9, is protective against ischemia-reperfusion (I/R) injury (Ref. 15). The mechanism, however, underlying this cardioprotection, is largely unknown. With evidence that activation of autophagy
Mahmood Khan et al.
Antioxidants & redox signaling, 11(4), 725-738 (2008-10-16)
The objective of this study was to establish the cardioprotective effect of sulfaphenazole (SPZ), a selective inhibitor of cytochrome P450 2C9 enzyme, in an in vivo rat model of acute myocardial infarction (MI). MI was induced by 30 min ligation
Vienna E Brunt et al.
The Journal of physiology, 590(15), 3523-3534 (2012-06-08)
While it is accepted that NO is responsible for ∼60% of the plateau in cutaneous thermal hyperaemia, a large portion of the response remains unknown. We sought to determine whether the remaining ∼40% could be attributed to EDHF-mediated activation of
Agostino Virdis et al.
The Journal of clinical endocrinology and metabolism, 95(2), 920-927 (2009-12-22)
The aim of this study was to assess whether patients with primary hyperparathyroidism (PHPT) show reduced endothelial function and to determine the mechanisms involved. The impact of parathyroidectomy (PTx) on endothelial function was also assessed. Endothelial dysfunction is reported in
Shu-Ying Chang et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(12), 2513-2522 (2008-09-13)
Various groups have sought to determine the impact of CYP2C8 genotype (and CYP2C8 inhibition) on the pharmacokinetics (PK) of ibuprofen (IBU) enantiomers. However, the contribution of cytochrome P450 2C8 (CYP2C8) in human liver microsomes (HLMs) has not been reported. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service