Skip to Content
Merck
All Photos(1)

Key Documents

31739

Supelco

Sulfisoxazole

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(3,4-dimethyl-5-isoxazolyl)benzenesulfonamide, N1-(3,4-Dimethyl-5-isoxazolyl)sulfanilamide, Sulfafurazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O3S
CAS Number:
Molecular Weight:
267.30
Beilstein:
6737262
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cc1noc(NS(=O)(=O)c2ccc(N)cc2)c1C

InChI

1S/C11H13N3O3S/c1-7-8(2)13-17-11(7)14-18(15,16)10-5-3-9(12)4-6-10/h3-6,14H,12H2,1-2H3

InChI key

NHUHCSRWZMLRLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfisoxazole belongs to the sulfonamide class of antibacterial compounds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfisoxazole may be used as a reference standard for the determination of sulfisoxazole in:
  • Honey after acidic hydrolysis by using post-column derivatization and high performance liquid chromatography (HPLC)-fluorescence detection.
  • Hospital, urban wastewater and river water by automated off-line solid phase extraction (SPE) followed by analysis using ultra-high-performance liquid chromatography combined with quadrupole linear ion trap tandem mass spectrometry (UHPLC-QqLIT).
  • Foodstuffs of animal origin by UHPLC coupled to tandem mass spectrometry (MS/MS).
  • Raw milk samples by SPE followed by UPLC equipped with time of flight mass spectrometry (TOF MS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Application of ion-exchange cartridge clean-up in food analysis: V. Simultaneous determination of sulphonamide antibacterials in animal liver and kidney using high-performance liquid chromatography with ultraviolet and mass spectrometric detection
Ito Y, et al.
Journal of Chromatography A, 898(1), 95-102 (2000)
Charles E Ahlfors et al.
Clinica chimica acta; international journal of clinical chemistry, 365(1-2), 78-85 (2005-09-20)
Measuring plasma unbound bilirubin concentration by the peroxidase test is useful in the management of jaundiced newborns. However, the commercially available peroxidase technology is manual, and the unbound bilirubin may be seriously underestimated at the 42-fold sample dilution and single
Melina Mondelli et al.
Journal of inorganic biochemistry, 102(2), 285-292 (2007-11-03)
The synthesis, structural characterization, voltammetric experiments and antibacterial activity of [Ni(sulfisoxazole)(2)(H(2)O)(4)].2H(2)O and [Ni(sulfapyridine)(2)] were studied and compared with similar previously reported copper complexes. [Ni(sulfisoxazole)(2)(H(2)O)(4)].2H(2)O crystallized in a monoclinic system, space group C2/c where the nickel ion was in a slightly
Rapid analysis of multiclass antibiotic residues and some of their metabolites in hospital, urban wastewater and river water by ultra-high-performance liquid chromatography coupled to quadrupole-linear ion trap tandem mass spectrometry
Gros M, et al.
Journal of Chromatography A, 1292(1), 173-188 (2013)
Hanne T Soligard et al.
Pediatric research, 67(6), 614-618 (2010-03-11)
Ibuprofen binds to plasma albumin and could interfere with the binding of bilirubin in jaundiced newborn infants. Most clinical studies have not shown increased concentrations of unbound bilirubin (UB) in plasma from infants treated with ibuprofen for a patent ductus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service