TRUEA
TrueGel3D Buffer pH 5.5
600 μl solution, 10X concentrated
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.21
Recommended Products
shipped in
dry ice
storage temp.
−20°C
General description
TrueGel3D buffer pH 5.5 is a component of the TrueGel3D Hydrogel system. The TrueGel3D buffer with pH 5.5 is recommended for fast gelling hydrogels, because the acidic pH (lower pH) slows down the formation of a gel allowing homogenous mixing of polymer and crosslinker before it solidify. However, in conjugation with TrueGel3D buffer pH 7.2, buffer with intermediate strengths can be prepared to modulate gelation kinetics. Both buffer systems (TRUEA and TRUEB) have pH indicator (phenol red) and strong buffering capacity to ensure a stable pH.
Application
TrueGel3D buffers with pH 5.5 and 7.2 is used to modulate gelation kinetics
Components
TRUEA-1VL contains TrueGel3D buffer pH 5.5, 10X concentrated
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service