Skip to Content
Merck
All Photos(1)

Key Documents

T7254

Sigma-Aldrich

Nα-Tosyl-L-lysine chloromethyl ketone hydrochloride

powder, ≥96% (TLC)

Synonym(s):

(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, (3S)-7-Amino-1-chloro-3-tosylamino-2-heptanone hydrochloride, TLCK, Tosyl-L-lysyl-chloromethane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21ClN2O3S · HCl
CAS Number:
Molecular Weight:
369.31
Beilstein:
7106867
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Product Name

Nα-Tosyl-L-lysine chloromethyl ketone hydrochloride, ≥96% (TLC), powder

biological source

synthetic (organic)

Assay

≥96% (TLC)

form

powder

potency

10-100 μM effective concentration

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

Cl[H].Cc1ccc(cc1)S(=O)(=O)N[C@@H](CCCCN)C(=O)CCl

InChI

1S/C14H21ClN2O3S.ClH/c1-11-5-7-12(8-6-11)21(19,20)17-13(14(18)10-15)4-2-3-9-16;/h5-8,13,17H,2-4,9-10,16H2,1H3;1H/t13-;/m0./s1

InChI key

YFCUZWYIPBUQBD-ZOWNYOTGSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Nα-Tosyl-L-lysine chloromethyl ketone hydrochloride has been used:
  • in chymotrypsin purification to prevent binding of trypsins to the affinity support by inhibiting them in the crude extract
  • as a protease inhibitor in tissue and cell homogenization
  • as a trypsin inhibitor to determine the specific protease activity levels

Biochem/physiol Actions

Nα-Tosyl-L-lysine chloromethyl ketone hydrochloride (TLCK) blocks the lipopolysaccharide (LPS)- or cytokine-induced activation of nuclear factor κB (NF-κB), which, in turn, blocks the induction of inducible nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) transcription. Blocks activation of pp70s6k by all mitogens. Blocks apoptosis in cell lines by inhibiting the processing of caspases in some cell lines and to some stimuli.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

Stable at least two years when stored desiccated at −20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F D'Acquisto et al.
FEBS letters, 418(1-2), 175-178 (1997-12-31)
We investigated the involvement of NF-kappaB in the regulation of COX-2 protein expression and prostaglandin production in LPS-stimulated J774 macrophages. Incubation of J774 cells with LPS (1 microg/ml) for 24 h caused an increase of COX-2 protein expression and accumulation
G Kwon et al.
Diabetes, 47(4), 583-591 (1998-05-06)
Interleukin-1beta (IL-1beta) has been implicated as an effector molecule of beta-cell destruction in autoimmune diabetes. IL-1beta inhibits insulin secretion from pancreatic beta-cells by stimulating the expression of inducible nitric oxide synthase (iNOS) that generates the free radical nitric oxide. IL-1beta
L Stefanis et al.
Journal of neurochemistry, 69(4), 1425-1437 (1997-11-05)
Rat pheochromocytoma (PC12) cells and sympathetic neurons undergo apoptotic cell death upon withdrawal of trophic support. We have shown previously that selective cysteine aspartase (caspase) inhibitors protect PC12 cells and sympathetic neurons from such death, and that the caspase Nedd-2
Feces derived allergens of Tyrophagus putrescentiae reared on dried dog food and evidence of the strong nutritional interaction between the mite and Bacillus cereus producing protease bacillolysins and exo-chitinases
Erban T, et al.
Frontiers in Physiology, 7, 53-53 (2016)
Purification and characterization of a serine protease with fibrinolytic activity from the dung beetles, Catharsius molossus
Ahn MY, et al.
Thrombosis Research, 112(5-6), 339-347 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service