Merck
  • Antioxidants, Enzyme Inhibitors, and Biogenic Compounds in Grain Extracts of Barleys.
Antioxidants, Enzyme Inhibitors, and Biogenic Compounds in Grain Extracts of Barleys.
Chemistry & biodiversity (2015-11-17)
Tibor Maliar, Gabriela Slaba, Peter Nemeček, Mária Maliarová, Michaela Benková, Michaela Havrlentová, Miroslav Ondrejovič, Ján Kraic
ABSTRACT
The content of biogenic compounds and the biological activities of barley (Hordeum vulgare L.)-grain extracts was evaluated. The sufficiently large and heterogeneous set of barley genotypes (100 accessions) enabled the selection of special genotypes interesting for potential industrial, pharmaceutical, and medicinal applications. Barley genotypes with the highest contents of phenols, phenolic acids, flavonoids, biogenic thiols, and amines, radical-scavenging activity, as well as inhibitory activities of trypsin, thrombin, collagenase, urokinase, and cyclooxygenase were identified.
Sigma-Aldrich
Trizma® hydrochloride, BioXtra, pH 3.5-5.0 (0.5 M in H2O), ≥99.0% (titration)
Sigma-Aldrich
Trizma® hydrochloride, BioPerformance Certified, suitable for cell culture, ≥99.0% (titration)
Sigma-Aldrich
Trizma® hydrochloride, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Trizma® hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline
Sigma-Aldrich
Z-L-Lys-SBzl hydrochloride