Skip to Content
Merck
All Photos(1)

Key Documents

T6664

Sigma-Aldrich

Tris Buffered Saline

powder, pH 8.0

Synonym(s):

TBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

product name

Tris Buffered Saline, pH 8.0, powder

form

powder

Quality Level

pH

8.0

Looking for similar products? Visit Product Comparison Guide

Application

Tris Buffered Saline has been used:
  • to block cells fixed in paraformaldehyde for immunofluorescence imaging
  • as a component in cell lysis buffer for DNA extraction
  • in goat serum and Triton-X 100 for blocking and permeabilizing cells for immunocytostaining respectively

Packaging

Foil pouches

Reconstitution

Contents of one pouch, when dissolved in one liter of distilled or deionized water, will yield 0.05 M Tris buffered saline (NaCl - 0.138 M; KCl - 0.0027 M); pH 8.0, at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin D Rynearson et al.
Bioorganic & medicinal chemistry, 28(22), 115734-115734 (2020-10-03)
The evolution of gamma-secretase modulators (GSMs) through the introduction of novel heterocycles with the goal of aligning activity for reducing the levels of Aβ42 and properties consistent with a drug-like molecule are described. The insertion of a methoxypyridine motif within
Arun Prasad Rao et al.
Contemporary clinical dentistry, 5(3), 296-301 (2014-09-06)
The development of dental caries has been associated with the oral prevalence of Streptococcus mutans. Four serotypes of S. mutans have been reported, namely serotype c, e, f, and k that are classified based on the composition and linkages of
Susanna Trapp et al.
Journal of virology, 83(2), 884-895 (2008-11-14)
Human immunodeficiency virus (HIV) is taken up by and replicates in immature dendritic cells (imDCs), which can then transfer virus to T cells, amplifying the infection. Strategies known to boost DC function were tested for their ability to overcome this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service