GE17-1321-01
Tris
Cytiva 17-1321-01, pack of 500 g
Synonym(s):
Tris Base, Tris Buffer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105323
NACRES:
NA.25
Recommended Products
form
solid
feature
wetted part: no
packaging
pack of 500 g
manufacturer/tradename
Cytiva 17-1321-01
color
White
pH range
7.2-9.0
solubility
cold water: soluble (Soluble in cold water and hot water)
methanol: soluble
storage temp.
room temp
Related Categories
General description
Tris (tris[hydroxymethyl] aminoethane) is a buffering agent commonly used in biochemical and molecular biology experiments to maintain a stable pH.
Features and Benefits
- Qualified for 2-D electrophoresis, Tris is suitable for preparing electrophoresis buffer in the pH range of 7.2 to 9.0.
- Tris is known for its ability to resist changes in pH when small amounts of acid or base are added, making it valuable in maintaining stable experimental conditions.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service