Skip to Content
Merck
All Photos(2)

Key Documents

T5783

Supelco

2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate

for chiral derivatization, LiChropur, ≥98.0% (HPLC)

Synonym(s):

GITC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO9S
CAS Number:
Molecular Weight:
389.38
Beilstein:
56699
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((chiral))
for chiral derivatization

Assay

≥98.0% (HPLC)

quality

LiChropur

technique(s)

HPLC: suitable

mp

114-116 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)OC[C@H]1O[C@@H](N=C=S)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C15H19NO9S/c1-7(17)21-5-11-12(22-8(2)18)13(23-9(3)19)14(24-10(4)20)15(25-11)16-6-26/h11-15H,5H2,1-4H3/t11-,12-,13+,14-,15-/m1/s1

InChI key

WOWNQYXIQWQJRJ-UXXRCYHCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate is a chiral derivatization reagent which reacts mainly with enantiomeric amino acids.

Application

Chiral reagent for resolution of amino acid derivatives.
GITC may have been used in the formation of diastereomers and for reversed-phase high-performance liquid chromatographic resolution of amino acid enantiomers.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L He et al.
Biomedical chromatography : BMC, 6(6), 291-294 (1992-11-01)
A high performance liquid chromatographic method was developed for the simultaneous assay of R(-)- and S(+)-albuterol in human serum. The assay involves solid phase extraction as a sample clean-up step and derivatization of racemic albuterol to its diastereomeric thioureas with
A Mozayani et al.
Journal of analytical toxicology, 19(6), 519-521 (1995-10-01)
The distribution of the racemic and the enantiomeric content of (+/-)-metoprolol was compared after ingestion of a massive fatal overdose of the racemic drug. Postmortem concentrations of the racemate in different tissues were assayed by gas chromatography after derivatization with
Reversed-phase liquid chromatographic resolution of amino acid enantiomers by derivatization with 2, 3, 4, 6-tetra-O-acetyl-?-D-glucopyranosyl isothiocyanate.
Nimura, Noriyuki, Haruo Ogura, and Toshio Kinoshita.
Journal of Chromatography A, 202.3, 375-379 (1980)
Bushra Saeed et al.
BMC genomics, 17, 98-98 (2016-02-06)
High potential of Morus laevigata and Morus serrata has been proposed in the breeding programs for Morus sp. However, due to the lack of dense molecular markers this goal is still in its nascent stage and not yet realized. We
S Chakravorty et al.
FEMS microbiology letters, 205(1), 113-117 (2001-12-01)
Nucleic acid amplification technologies offer great promise for the rapid, sensitive and specific diagnosis of tuberculosis. However, the isolation of inhibitor-free DNA from biological specimens is a bottleneck of the PCR assay. Here we describe a simple method for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service