Skip to Content
Merck
All Photos(1)

Key Documents

T5513

Sigma-Aldrich

N,N,N′,N′-Tetramethylbenzidine

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-C6H4-4-N(CH3)2]2
CAS Number:
Molecular Weight:
240.34
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95% (HPLC)

form

powder

mp

193-195 °C (lit.)

SMILES string

CN(C)c1ccc(cc1)-c2ccc(cc2)N(C)C

InChI

1S/C16H20N2/c1-17(2)15-9-5-13(6-10-15)14-7-11-16(12-8-14)18(3)4/h5-12H,1-4H3

InChI key

YRNWIFYIFSBPAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Stürmer et al.
Neuroscience letters, 23(1), 1-5 (1981-04-09)
Tetramethylbenzidine (TMB) histochemistry for retrogradely transported horseradish peroxidase (HRP) has been successfully applied in electron microscopical preparations in the guinea pig visual cortex. In ultrathin sections, the TMB--HRP reaction product appears as an accumulation of typical electron-dense crystals. These crystals
T M Bricker et al.
Biochimica et biophysica acta, 1409(1), 50-57 (1998-11-07)
Site-directed mutagenesis was used to produce a Synechocystis mutant containing a histidine tag at the C terminus of the CP 47 protein of Photosystem II. This mutant cell line, designated HT-3, exhibited slightly above normal rates of oxygen evolution and
L G McGirr et al.
Chemico-biological interactions, 61(1), 61-74 (1987-01-01)
The mechanism of peroxidative N-dealkylation of alkylamines proceeds via one-electron oxidation to the iminium cation which reacts with water to give the N-hydroxymethyl derivative which decomposes to formaldehyde and the N-demethylated product. This reaction is normally inhibited by glutathione by
M Kalia et al.
Brain research, 188(1), 23-32 (1980-04-21)
The intra-axonal transport of horseradish peroxidase (HRP) has been used to trace the nodose ganglion and brain stem projections of a physiologically distinct nerve - the aortic depressor nerve - following electrophysiological identification. Tetramethyl benzidine (TMB) has been used as
S Kusunoki et al.
Journal of clinical microbiology, 29(8), 1596-1603 (1991-08-01)
Quantitative microdilution plate hybridization was used to identify 22 Mycobacterium species. DNAs of clinical strains were rapidly extracted and labeled with photoreactive biotin. Labeled DNAs were distributed into wells of a microdilution plate in which reference DNAs had been immobilized.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service