Skip to Content
Merck
All Photos(1)

Key Documents

T3651

Sigma-Aldrich

Anti-Tropomyosin antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

chicken

technique(s)

indirect immunofluorescence: 1:40 using cultured chicken fibroblasts
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

chicken ... TPM1(396366)

General description

Tropomyosin is a rigid, rod shaped protein, closely associated with actin filaments. In non-muscle cells grown in culture, tropomyosin is distributed periodically along the length of the stress fibers.

Specificity

The antibody specifically reacts with tropomyosin (typical banded pattern of stress fibers) in cultured cells and two tropomyosin bands of total chicken gizzard extract by immunoblotting.

Immunogen

smooth muscle tropomyosin from chicken gizzard.

Application

Anti-Tropomyosin antibody produced in rabbit has been used in western immunoblotting and immunofluorescence.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Miyado et al.
Biochemical and biophysical research communications, 225(2), 427-435 (1996-08-14)
Tropomyosin is an actin-associated cytoskeletal protein expressed in muscle and non-muscle cells. There are several tropomyosin isoforms, and their cellular expression is known to be associated with transformation events caused by retroviral infection and chemical mutagens. We found that expression
Differentiation of CD45?/CD31+ lung side population cells into endothelial and smooth muscle cells in vitro
Xu Y, et al.
International Journal of Molecular Medicine, 43(3), 1128-1138 (2019)
Fang Wang et al.
Journal of biomedicine & biotechnology, 2011, 435271-435271 (2011-12-22)
Familial hypertrophic cardiomyopathy (FHC) is a disease of cardiac sarcomeres. To identify molecular mechanisms underlying FHC pathology, functional and structural differences in three FHC-related mutations in recombinant α-Tm (V95A, D175N, and E180G) were characterized using both conventional and modified in
In vitro proteolysis of myofibrillar and sarcoplasmic proteins of European sea bass (Dicentrarchus Labrax L) by an endogenous m-calpain.
Veronique Verrez-Bagnis
Journal of the Science of Food and Agriculture, 82(11), 1256-1262 (2002)
Yang Xu et al.
International journal of molecular medicine, 43(3), 1128-1138 (2019-01-11)
Side population (SP) cells are a small subpopulation of cells found in many mammalian tissues and organs, identified by their capacity to efflux Hoechst 33342 dye. They are enriched for stem/progenitor cell activity. SP cells isolated from the adult mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service