Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200715

Sigma-Aldrich

Anti-β-tubulin antibody, Mouse monoclonal

clone 2-28-33, purified from hybridoma cell culture

Synonym(s):

Anti-TUBB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

2-28-33, monoclonal

form

buffered aqueous solution

mol wt

~50 kDa

species reactivity

dog, chicken, rat, bovine, monkey, human, mouse

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 0.5 μg/mL using mouse embryo fibroblast NIH-3T3 cells extract
immunofluorescence: 1-2 μg/mL using human foreskin fibroblast HS68 cells
immunohistochemistry: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBB(203068)
mouse ... Tubb3(22152)

General description

Anti-β-tubulin antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the 2-28-33 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Tubulin also known as TUBB is the major building block of microtubules. Microtubules assemble from heterodimers of α-tubulin and β-tubulin; both have a molecular weight of ~ 50 kDa and share considerable homology. For β-tubulin, five evolutionarily conserved isotypes have been identified, which are regularly utilized in the same cell types of different species.

Immunogen

Sarkosyl-resistant ribbons from Strongylocentrotus purpuratus (sea urchin) sperm axonemes.

Application

Monoclonal Anti-β-tubulin has been used:
  • in immunoblotting
  • in immunofluorescence
  • in immunohistochemistry

Biochem/physiol Actions

Monoclonal Anti-β-tubulin specifically recognizes the two major β-tubulin isotypes and one of the minor β-tubulin isotypes of the free-living soil nematode Caenorhabditis elegans. It also binds to Sarkosyl resistant ribbons from Strongylocentrotus purpuratus sperm axonemes, neuronal axons of the larva of Ciona intestinalis (ascidian), microtubules in root tip cells in Arabidopsis thaliana and to human, mouse, rat, monkey, bovine, dog and chicken β-tubulin. The different β-tubulin isotypes are suggested to differ from each other in their ability to polymerize into microtubules.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Icariin Attenuates High Glucose-Induced Apoptosis, Oxidative Stress, and Inflammation in Human Umbilical Venous Endothelial Cells
Sun S, et al.
Planta Medica, 85(06), 473-482 (2019)
Naoki Goto et al.
Developmental neuroscience, 45(1), 19-26 (2023-01-12)
Rho family small GTPases, such as Rho, Rac, and Cdc42, play essential roles during brain development, by regulating cellular signaling and actin cytoskeletal reorganization. Rich2/Arhgap44, a Rac- and Cdc42-specific GTPase-activating protein, has been reported to be a key regulator for
Xinghua Zhang et al.
Journal of thoracic disease, 12(10), 5887-5895 (2020-11-20)
Little is known about the relationship between ubiquilin 1 (UBQLN1) and p53, both of them have been implicated in the development and progression of non-small cell lung cancer (NSCLC). In this study, we aimed to explore the role of loss
Xiaohua Ma et al.
Journal of cellular and molecular medicine, 25(24), 11300-11309 (2021-11-21)
βII spectrin (β2SP) is encoded by Sptbn1 and is involved in the regulation of various cell functions. β2SP contributes to the formation of the myelin sheath, which may be related to the mechanism of neuropathy caused by demyelination. As one
Bin Zuo et al.
Cellular and molecular gastroenterology and hepatology, 17(6), 1039-1061 (2024-03-12)
The functional maturation of the liver largely occurs after birth. In the early stages of life, the liver of a newborn encounters enormous high-fat metabolic stress caused by the consumption of breast milk. It is unclear how the maturing liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service