Skip to Content
Merck
All Photos(1)

Key Documents

T3452

Sigma-Aldrich

Toloxatone

≥98% (HPLC), solid

Synonym(s):

5-(Hydroxymethyl)-3-(3-methylphenyl)-2-oxazolidinone, Humoryl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO3
CAS Number:
Molecular Weight:
207.23
EC Number:
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Cc1cccc(c1)N2CC(CO)OC2=O

InChI

1S/C11H13NO3/c1-8-3-2-4-9(5-8)12-6-10(7-13)15-11(12)14/h2-5,10,13H,6-7H2,1H3

InChI key

MXUNKHLAEDCYJL-UHFFFAOYSA-N

Biochem/physiol Actions

Toloxatone is a reversible monoamine oxidase A inhibitor (MAOI) and antidepressant. Toloxatone belongs to the aryloxazolidinones, a relatively new class of reversible monoamine oxidase A inhibitors (MAOI). Studies show that derivatives such as 3-aryloxazolidin-2-one (oxazolidinones), which are regularly used to treat gram positive infections, represent a new class of reversible as well as irreversible inhibitors for MAO-A and MAO-B enzymes that are able to inhibit protein synthesis and also used for neurodegenerative-type pathologies.

Preparation Note

Toloxatone is soluble in DMSO at a concentration that is greater than 10 mg/ml.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geum Seok Jeong et al.
Molecules (Basel, Switzerland), 25(17) (2020-08-30)
Eight compounds were isolated from the roots of Glycyrrhiza uralensis and tested for cholinesterase (ChE) and monoamine oxidase (MAO) inhibitory activities. The coumarin glycyrol (GC) effectively inhibited butyrylcholinesterase (BChE) and acetylcholinesterase (AChE) with IC50 values of 7.22 and 14.77 µM
P Lemoine et al.
Psychopharmacology, 106 Suppl, S118-S119 (1992-01-01)
A double-blind comparison of moclobemide and toloxatone was performed in adult out-patients diagnosed as suffering from a major depressive disorder. Parallel groups of patients received moclobemide, 450 mg/day (n = 135) or toloxatone, 1000 mg/day (n = 133) for 28
Seung Cheol Baek et al.
Bioorganic chemistry, 83, 317-325 (2018-11-06)
Three flavanones and two flavones were isolated from the leaves of Prunus padus var. seoulensis by the activity-guided screening for new monoamine oxidase (MAO) inhibitors. Among the compounds isolated, rhamnocitrin (5) was found to potently and selectively inhibit human MAO-A
M L Kaltenbach et al.
Alcoholism, clinical and experimental research, 23(5), 885-890 (1999-06-17)
The pharmacokinetics of toloxatone and ethanol were determined in plasma and cerebrospinal fluid of conscious rabbits. According to a cross-over design, rabbits (n = 5) randomly received on three separate occasions either toloxatone (5 mg/kg), ethanol (1 g/kg), or toloxatone
R Vistelle et al.
Journal of chromatography, 490(2), 387-394 (1989-05-30)
A simplified, rapid high-performance liquid chromatographic procedure has been developed for the measurement of toloxatone in rabbit plasma and cerebrospinal fluid. The method involves a single-step extraction of the alkalinized sample with diethyl ether and analysis of the evaporated extract

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service