Skip to Content
Merck
All Photos(1)

Key Documents

T3397

Sigma-Aldrich

Tylosin solution

solution, suitable for cell culture, BioReagent

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.76

Product Name

Tylosin solution, 8 mg/mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture

Quality Level

sterility

0.1 μm filtered

product line

BioReagent

form

solution

concentration

8 mg/mL in 0.9% NaCl

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

colorless to yellow

antibiotic activity spectrum

Gram-positive bacteria
mycoplasma

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.CC[C@H]1OC(=O)C[C@@H](O)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](O)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CC=O)C[C@@H](C)C(=O)\C=C\C(C)=C\[C@@H]1CO[C@@H]4O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]4OC

InChI

1S/C46H77NO17.C4H6O6/c1-13-33-30(22-58-45-42(57-12)41(56-11)37(52)26(5)60-45)18-23(2)14-15-31(49)24(3)19-29(16-17-48)39(25(4)32(50)20-34(51)62-33)64-44-38(53)36(47(9)10)40(27(6)61-44)63-35-21-46(8,55)43(54)28(7)59-35;5-1(3(7)8)2(6)4(9)10/h14-15,17-18,24-30,32-33,35-45,50,52-55H,13,16,19-22H2,1-12H3;1-2,5-6H,(H,7,8)(H,9,10)/b15-14+,23-18+;/t24-,25+,26-,27-,28+,29+,30-,32-,33-,35+,36-,37-,38-,39-,40-,41-,42-,43+,44+,45-,46-;1-,2-/m11/s1

InChI key

ICVKYYINQHWDLM-KBEWXLTPSA-N

General description

Chemical structure: macrolide
Tylosin is a macrolide antibiotic, which comprises of a 16-member lactone ring. This solution is recommended for use at 1 mL/L.

Application

Tylosin solution has been used as an antimicrobial agent in antibiotic susceptibility minimal inhibitory concentration(MIC) of E. coli and S. aureus isolates. It has also been used as a component of Dulbecco′s modified Eagle′s (DMEM) medium for self-inactivating lentivirus production and skin fibroblast cultivation.

Biochem/physiol Actions

Tylosin inhibits bacterial protein synthesis by binding to the large subunit 50S. It is effective against Gram-positive bacteria and mycoplasma and is used in veterinary medicine. It has been used to study implant site abscess prevention in cattle and Mycoplasma gallisepticum and Mycoplasma meleagridis infections.

Packaging

20ML

Preparation Note

This product was formulated to contain 8 mg/ml tylosin in 0.9% sodium chloride.

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and keep upright to prevent leakage.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of a tylosin tartrate pellet added to cattle growth implants on the incidence of implant site abscesses
Zollers JrWG, et al.
The Protein Kinase Facts Book, 18(3), 258-261 (2002)
Tools for characterizing bacterial protein synthesis inhibitors
Orelle C, et al.
Antimicrobial Agents and Chemotherapy, 57(12), 5994-6004 (2013)
Antibiotic susceptibility patterns of beta-lactamase-producing Escherichia coli and Staphylococcus aureus isolated from chickens in Maiduguri (Arid zone), Nigeria
A Mamza S, et al.
Veterinarski Archiv / Veterinary Archives, 80(2), 283-297 (2010)
EVALUATION OF THE EFFECT OF TYLOSIN (MACROLIDE ANTIBIOTICS) ON MURINE TOXOPLASMOSIS
Asgari Q, et al.
International Journal of Pharmaceutical Sciences and Research, 9(7), 2891-2896 (2018)
Frizzled 1 and Wnt1 as new potential therapeutic targets in the traumatically injured spinal cord
Gonzalez P, et al.
Cellular and Molecular Life Sciences, 80, 1-32 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service