Skip to Content
Merck
All Photos(1)

Key Documents

T2694

Sigma-Aldrich

Trizma® hydrochloride solution

pH 8.0, BioPerformance Certified, 1 M, suitable for cell culture

Synonym(s):

Tris hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

grade

BioPerformance Certified
Molecular Biology
for molecular biology

Quality Level

form

solution

concentration

1 M

technique(s)

cell culture | mammalian: suitable

impurities

DNase, RNase, Protease, none detected
bioburden, tested
endotoxin, tested
≤5 ppm Heavy metals (as Pb)

pH

8.0

useful pH range

7.0-9.0

absorption

≤0.05 at 290 at 40%

Looking for similar products? Visit Product Comparison Guide

General description

A series of Pre-mixed solutions of TRIZMA Base and TRIZMA HCl to provide commonly used pH values for Tris buffers. No mixing or pH adjustment necessary. Guaranteed accuracy ± 0.1 pH units.

Application

Trizma® hydrochloride solution has been used:
  • as a component of extraction buffer for Plasmodiophora brassicae DNA isolation
  • as a component of resuspension buffer for human genomic DNA fragmented sample
  • for adjusting pH of plasma samples and as a component of reconstitution and neutralization buffer

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Accurate variant detection across non-amplified and whole genome amplified DNA using targeted next generation sequencing
ElSharawy A, et al.
BMC Genomics, 13(1), 500-500 (2012)
Anti-peptide monoclonal antibodies generated for immuno-MRM assays have a high probability of supporting Western blot and ELISA
Schoenherr RM, et al.
Molecular and Cellular Proteomics, mcp-O114 (2014)
Olivier Harismendy et al.
Genome biology, 12(12), R124-R124 (2011-12-22)
Ultra-deep targeted sequencing (UDT-Seq) can identify subclonal somatic mutations in tumor samples. Early assays' limited breadth and depth restrict their clinical utility. Here, we target 71 kb of mutational hotspots in 42 cancer genes. We present novel methods enhancing both
Root Gall Formation, Resting Spore Isolation and High Molecular Weight DNA Extraction of Plasmodiophora brassicae
Mehrabi S, et al.
Bio-protocol (2018)
Markus Frederik Schliffka et al.
eLife, 10 (2021-04-20)
During the first days of mammalian development, the embryo forms the blastocyst, the structure responsible for implanting the mammalian embryo. Consisting of an epithelium enveloping the pluripotent inner cell mass and a fluid-filled lumen, the blastocyst results from a series

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service