Skip to Content
Merck
All Photos(1)

Key Documents

T2569

Sigma-Aldrich

Trizma® hydrochloride solution

pH 7.8, BioPerformance Certified, 1 M, suitable for cell culture

Synonym(s):

Tris hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

grade

BioPerformance Certified
Molecular Biology
for molecular biology

Quality Level

form

solution

concentration

1 M

technique(s)

cell culture | mammalian: suitable

impurities

DNase, RNase, Protease, none detected
bioburden, tested
endotoxin, tested
≤5 ppm Heavy metals (as Pb)

pH

7.8

useful pH range

7.0-9.0

absorption

≤0.05 at 290 at 40%

Looking for similar products? Visit Product Comparison Guide

General description

A series of Pre-mixed solutions of TRIZMA Base and TRIZMA HCl to provide commonly used pH values for Tris buffers. No mixing or pH adjustment necessary. Guaranteed accuracy ± 0.1 pH units.
Tris(hydroxymethyl)aminomethane (Tris) is a routine buffer in biological sciences. The pH buffering range is between 7.0 -9.0. It displays negligible binding towards metal in solutions. Tris is used majorly in combination with ethylenediaminetetraacetic acid (EDTA), especially for electrophoretic studies. Its low ionic mobility finds usage in capillary electrochromatography as well.

Application

Trizma® hydrochloride solution has been used:
  • as a component of lysis buffer for lysing human embryonic kidney (HEK) 293 cells
  • in the 10X digoxigenin (DIG) deoxynucleoside triphosphate (dNTP) nick translation labeling buffer and in tris EDTA and sodium chloride (TEN) buffer
  • in the preparation of tris, acetate, ethylenediaminetetraacetic acid (EDTA) (1x TAE) buffer and as a component of 10X gel loading dye

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vaccine technologies for veterinary viral diseases null
Universal buffers for use in biochemistry and biophysical experiments
Brooke D, et al.
AIMS Biophysics, 2(3) (2)
A review of acellular immersion tests on bioactive glasses-influence of medium on ion release and apatite formation
Nommeots-Nomm A, et al.
International Journal of Applied Glass Science, 11(3) (11)
Stuart W Tompson et al.
Bio-protocol, 7(10) (2017-08-02)
There are several in silico programs that endeavor to predict the functional impact of an individual's sequence variation at splice donor/acceptor sites, but experimental confirmation is problematic without a source of RNA from the individual that carries the variant. With
(Un) suitability of the use of pH buffers in biological, biochemical and environmental studies and their interaction with metal ions-a review
Ferreira CMH, et al.
Royal Society of Chemistry Advances, 5(39) (5)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service