Skip to Content
Merck
All Photos(1)

Key Documents

T2144

Sigma-Aldrich

N,N′,N′′-Triacetylchitotriose

≥93% (HPLC)

Synonym(s):

NAG3, Tri(N-acetyl-D-glucosamine), O-[2-Acetamido-2-deoxy-β-D-glucopyranosyl-(1→4)]-O-[2-acetamido-2-deoxy-β-D-glucopyranosyl-(1→4)]-2-acetamido-2-deoxy-D-glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H41N3O16
CAS Number:
Molecular Weight:
627.59
Beilstein:
75249
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥93% (HPLC)

form

powder

impurities

≤8% water (Karl Fischer)

color

white to yellow cast

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H](C=O)[C@@H](O)[C@H](O[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2NC(C)=O)[C@H](O)[C@H]1NC(C)=O)[C@H](O)CO

InChI

1S/C24H41N3O16/c1-8(32)25-11(4-28)17(36)21(12(35)5-29)42-24-16(27-10(3)34)20(39)22(14(7-31)41-24)43-23-15(26-9(2)33)19(38)18(37)13(6-30)40-23/h4,11-24,29-31,35-39H,5-7H2,1-3H3,(H,25,32)(H,26,33)(H,27,34)/t11-,12+,13+,14+,15+,16+,17+,18+,19+,20+,21+,22+,23-,24-/m0/s1

InChI key

LRDDKCYRFNJZBX-WHFMPQCRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N′,N′′-Triacetylchitotriose has been used:
  • as a competitive inhibitor of the enzymatic activity of lysozyme to treat Macrophages to inhibit the constitutively lysozyme production[1]
  • to dock the catalytic cleft of Cel9A (endocellulases)[2]
  • as an inhibitory sugar to access the specificity of labelling with gold complexed lectin[3]

Biochem/physiol Actions

N,N′,N′′-Triacetylchitotriose is a chitin oligosaccharide,[4] that promotes lysozyme inhibition.[5] Chitin oligosaccharides are associated with many physiological functions. They are biodegradable and are useful in identifying chitin polysaccharides and associated glycoconjugates.[4]

Preparation Note

Prepared by the method of Barker, S.A., et al., J. Chem. Soc., 2218 (1958).

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysozyme plays a dual role against the dimorphic fungus Paracoccidioides brasiliensis
Lopera D, et al.
Revista Do Instituto De Medicina Tropical De Sao Paulo, 50(3), 169-175 (2008)
Chitin binding by Thermobifida fusca cellulase catalytic domains
Li Y and Wilson DB
Biotechnology and Bioengineering, 100(4), 644-652 (2008)
Analysis of chitin oligosaccharides by capillary electrophoresis with laser-induced fluorescence
Wang CY and Hsieh YZ
Journal of Chromatography A, 979(1-2), 431-438 (2002)
Isolation and immunolocalization of a Pinus nigra lectin (PNL) during interaction with the necrotrophs?Heterobasidion annosum and Fusarium avenaceum
Nahalkova J, et al.
Physiological and Molecular Plant Pathology, 59(3), 153-163 (2001)
Staphylococcus aureus Evades Lysozyme-Based Peptidoglycan Digestion that Links Phagocytosis, Inflammasome Activation, and IL-1? Secretion
Shimada T, et al.
Cell host & microbe, 7(1), 38-49 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service