Skip to Content
Merck
All Photos(1)

Key Documents

T133

Sigma-Aldrich

Inactin® hydrate

≥98% (HPLC)

Synonym(s):

Thiobutabarbital sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N2NaO2S · xH2O
CAS Number:
Molecular Weight:
250.29 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

color

light yellow

solubility

H2O: soluble (storage of solutions for more than 8 hours at 4°C is not recommended.)

SMILES string

[Na+].CCC(C)C1(CC)C([O-])=NC(=S)NC1=O

InChI

1S/C10H16N2O2S.Na/c1-4-6(3)10(5-2)7(13)11-9(15)12-8(10)14;/h6H,4-5H2,1-3H3,(H2,11,12,13,14,15);/q;+1/p-1

InChI key

SLZHLQUFNFXTHB-UHFFFAOYSA-M

Application

Inactin® hydrate has been used to anesthetize rats to study stress-induced hypersensitivity and mice to study Cisplatin-induced neuropathy. It has also been used to anesthetize rats to measure the glomerular filtration rate (GFR).

Biochem/physiol Actions

Inactin® is a long-lasting rodent anesthetic with minimal effects on cardiovascular tone and renal output. It exhibits sedative and hypnotic properties.

Legal Information

Inactin is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flecknell
Laboratory Animal Anesthesia. An introduction for Research Workers and Technicians, 36-36 (1987)
Min-Suk Yoon et al.
BMC neuroscience, 10, 77-77 (2009-07-16)
Cisplatin mediates its antineoplastic activity by formation of distinct DNA intrastrand cross links. The clinical efficacy and desirable dose escalations of cisplatin are restricted by the accumulation of DNA lesions in dorsal root ganglion (DRG) cells leading to sensory polyneuropathy
Maki Niihori et al.
American journal of respiratory cell and molecular biology, 62(2), 231-242 (2019-08-29)
NFU1 is a mitochondrial protein that is involved in the biosynthesis of iron-sulfur clusters, and its genetic modification is associated with disorders of mitochondrial energy metabolism. Patients with autosomal-recessive inheritance of the NFU1 mutation G208C have reduced activity of the
Kevin T Bush et al.
JCI insight, 5(7) (2020-04-10)
The role of the renal organic anion transporters OAT1 (also known as SLC22A6, originally identified as NKT) and OAT3 (also known as SLC22A8) in chronic kidney disease (CKD) remains poorly understood. This is particularly so from the viewpoint of residual
Eman Y Gohar et al.
Journal of the American Heart Association, 9(10), e015110-e015110 (2020-05-12)
Background The novel estrogen receptor, G-protein-coupled estrogen receptor (GPER), is responsible for rapid estrogen signaling. GPER activation elicits cardiovascular and nephroprotective effects against salt-induced complications, yet there is no direct evidence for GPER control of renal Na+ handling. We hypothesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service