T1162
Tetramethylrhodamine isothiocyanate–Dextran
average mol wt 65,000-85,000
Synonym(s):
TRITC-Dextran
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder
mol wt
average mol wt 65,000-85,000
extent of labeling
~1 TRITC per 100 glucose residues
color
purple to pink
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Tetramethylrhodamine isothiocyanate-Dextran/TRITC-Dextran is a fluorochrome.
Application
Tetramethylrhodamine isothiocyanate-Dextran has been used to evaluate the role of pores (and channels) in the initial stages of amylolysis. It has also been used in protein-fluorescent conjugation.
Other Notes
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Amylase binding to starch granules under hydrolysing and non-hydrolysing conditions
Dhital S, et al.
Carbohydrate Polymers, 113, 97-107 (2014)
Evaluation of fluorescent plasma markers for in vivo microscopy of the microcirculation
Reeves KJ, et al.
Journal of Vascular Research, 49(2), 132-143 (2012)
Stephen Sakuma et al.
Cancer discovery, 11(1), 176-193 (2020-09-30)
Nuclear pore complexes (NPC) are the central mediators of nucleocytoplasmic transport. Increasing evidence shows that many cancer cells have increased numbers of NPCs and become addicted to the nuclear transport machinery. How reducing NPC numbers affects the physiology of normal
Nicholas E Propson et al.
The Journal of clinical investigation, 131(1) (2020-09-30)
Dysfunction of immune and vascular systems has been implicated in aging and Alzheimer disease; however, their interrelatedness remains poorly understood. The complement pathway is a well-established regulator of innate immunity in the brain. Here, we report robust age-dependent increases in
Hematopoietic Cancers
Immune Response, 1025-1063 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service