Skip to Content
Merck
All Photos(1)

Documents

T0895

Sigma-Aldrich

Thymine

suitable for cell culture, BioReagent

Synonym(s):

2,4-Dihydroxy-5-methylpyrimidine, 5-Methyluracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
Beilstein:
117880
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic

product line

BioReagent

Assay

≥99% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

mp

~320 °C (dec.) (lit.)

solubility

1 M NaOH: 50 mg/mL, clear to hazy, colorless to faintly yellow

SMILES string

CC1=CNC(=O)NC1=O

InChI

1S/C5H6N2O2/c1-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)

InChI key

RWQNBRDOKXIBIV-UHFFFAOYSA-N

Gene Information

mouse ... Tymp(72962)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefano Stella et al.
Acta crystallographica. Section D, Biological crystallography, 69(Pt 9), 1707-1716 (2013-09-04)
Transcription activator-like effectors contain a DNA-binding domain organized in tandem repeats. The repeats include two adjacent residues known as the repeat variable di-residue, which recognize a single base pair, establishing a direct code between the dipeptides and the target DNA.
Subhendu Sekhar Bag et al.
The Journal of organic chemistry, 78(2), 278-291 (2012-11-23)
We report the design and synthesis of triazolyl donor/acceptor unnatural nucleosides via click chemistry and studies on the duplex stabilization of DNA containing two such new nucleosides. The observed duplex stabilization among the self-pair/heteropair has been found to be comparable
Javier A Tello et al.
American journal of physiology. Endocrinology and metabolism, 305(1), E67-E77 (2013-05-02)
The human GnRH receptor (GNRHR1) has a specific set of properties with physiological and pharmacological influences not appropriately modeled in laboratory animals or cell-based systems. To address this deficiency, we have generated human GNRHR1 knock-in mice and described their reproductive
S I Ahmad et al.
Annual review of microbiology, 52, 591-625 (1999-01-19)
For many years it has been known that thymine auxotrophic microorganisms undergo cell death in response to thymine starvation [thymineless death (TLD)]. This effect is unusual in that deprivation of many other nutritional requirements has a biostatic, but not lethal
M Weinfeld et al.
Progress in nucleic acid research and molecular biology, 68, 139-149 (2001-09-14)
The toxic and mutagenic effects of ionizing radiation are believed to be caused by damage to cellular DNA. We have made use of a novel immunoassay for thymine glycol to examine the removal of this lesion from the DNA of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service