SRP6340
IL18BP human
recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Synonym(s):
IL18BPa, Interleukin-18-binding protein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥90% (SDS-PAGE)
form
liquid
mol wt
20 kDa (187 aa, 31-194 aa + His Tag)
packaging
pkg of 100 μg
concentration
1 mg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... IL18BP(10068)
General description
IL18BP functions as an inhibitor of the proinflammatory cytokine, IL18. It binds IL18, prevents the binding of IL18 to its receptor, and thus inhibits IL18-induced IFN-gamma production, resulting in reduced T-helper type 1 immune responses. This protein is constitutively expressed and secreted in mononuclear cells. Elevated level of this protein is detected in the intestinal tissues of patients with Crohn′s disease. Recombinant human IL18BP protein, fused to His-tag at N-terminus, was expressed in E.coli.
Physical form
1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.4 M Urea and 10% glycerol.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ahmet Bal et al.
The Journal of surgical research, 201(1), 13-21 (2016-02-07)
The aim of this study was to investigate the possible protective effect of interleukin 18-binding protein (IL-18BP) on ischemia-reperfusion (I/R)-induced liver injury in experimental rat models. Liver is one of the most affected organs from I/R process. IL-18 is an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service