Skip to Content
Merck
All Photos(1)

Key Documents

RAB0543

Sigma-Aldrich

Human IL18 / Interleukin-18 ELISA Kit

for serum, plasma, cell culture supernatants and urine

Synonym(s):

IFN-gamma-inducing factor, IL-18, Interleukin-18 

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type urine
sample type plasma
sample type serum

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.5 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL18(3606)

General description

The antibody pair provided in this kit detects Human IL-18 in serum, plasma, cell culture supernatants, and urine. Interleukin-18 (IL-18) is a proinflammatory cytokine, expressed in most of the mammalian tissues including adipose, liver and skeletal muscle. It is generated as pro-IL-18 which undergoes cleavage to form a mature IL-18 possessing the signal peptide. IL-18 induces Th1 (T helper cell 1) lymphocyte action mediated by interferon-γ. It controls the metabolism of lipids and glucose. Inflammasome activation is responsible for IL-18 release, which is a significant event in response to viral infection. Increased level of IL-18 is observed in kidney and liver disease, diabetes, atherosclerosis, autoimmune disease such as rheumatoid arthritis and asthma. It is also associated with the development of renal dysfunction.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human IL18 / Interleukin-18 ELISA Kit has been used to evaluate the macrophage cytokines production in response to trans-stilbene and terphenyl compounds. It has also been used to analyze and measure IL-18 levels in the urine of chronic kidney disease patients.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Victor Edgar Fiestas Solórzano et al.
Viruses, 13(9) (2021-09-29)
The incidence of dengue in Latin America has increased dramatically during the last decade. Understanding the pathogenic mechanisms in dengue is crucial for the identification of biomarkers for the triage of patients. We aimed to characterize the profile of cytokines
Quantification of Inflammasome Adaptor Protein ASC in Biological Samples by Multiple-Reaction Monitoring Mass Spectrometry
Ulke-Lemee A, et al.
Inflammation, 41(4), 1396-1408 (2018)
Effects of trans-stilbene and terphenyl compounds on different strains of Leishmania and on cytokines production from infected macrophages
Bruno F, et al.
Experimental Parasitology, 184(1), 31-38 (2018)
Yi-Chao Chai et al.
World journal of gastroenterology, 23(40), 7221-7231 (2017-11-17)
To compare the effect of University of Wisconsin (UW) solution with or without metformin, an AMP-activated protein kinase (AMPK) activator, for preserving standard and marginal liver grafts of young and aged rats Eighteen young (4 mo old) and 18 aged
Systemic Elevation of Proinflammatory Interleukin-18 in Human Immunodeficiency Virus (HIV)/Hepatitis C Virus (HCV) Coinfection Versus HIV or HCV Monoinfection
Veenhuis RT, et al.
Clinical Infectious Diseases, 64(5), 589-596 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service