Skip to Content
Merck
All Photos(1)

Documents

SRP3276

Sigma-Aldrich

SDF-1 ALPHA human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CXCL12, PBSF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

20.0-80.0 ng/mL

mol wt

8.0 kDa

packaging

pkg of 10 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CXCL12(6387)

General description

SDF-1α (stromal cell-derived factor 1 α) and β are stromal derived CXC chemokines, and signal through the CXCR4 (C-X-C motif chemokine receptor). SDF1α is an isoform of CXCL12 (C-X-C motif chemokine 12, also referred to as SDF-1). The gene is mapped to human chromosome 10q11. Recombinant human SDF-1a is an 8.0kDa protein containing 68 amino acid residues.

Application

SDF-1α (stromal cell-derived factor 1 α) human has been used to study the role of SDF-1α in cell migration and survival after TGF (transforming growth factor)-β-induced EMT (epithelial–mesenchymal transition) in hepatoma cells. It has been used to study podia formation, migration, proliferation and cell-cell adhesion of human hematopoietic progenitor cells in presence of SDF-1α.
SDF-1α (stromal cell-derived factor 1 α) human has been used in the chemotaxis assay of neural crest cells (NCCs) and as a component of Dulbecco′s modified Eagle′s medium (DMEM) in migration assay of human mesenchymal stem cells (hMSCs).

Biochem/physiol Actions

SDF-1α (stromal cell-derived factor 1 α) and β chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. SDF-1 is mainly expressed in the inflamed and failing myocardium. It is involved in inflammation, proliferation, tumorigenicity and metastasis. The SDF-1 proteins exert HIV (human immunodeficiency virus) suppressive activity in cells expressing the CXCR4 (C-X-C motif chemokine receptor 4). It is upregulated in breast carcinoma, ovarian carcinoma and papillary thyroid carcinoma. It enhances angiogenesis in the ischemic tissue. SDF1 is also involved in neuroprotection after ischemic stroke with the help of CXCR4.

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The chemokine SDF-1 is a chemoattractant for human CD34+ hematopoietic progenitor cells and provides a new mechanism to explain the mobilization of CD34+ progenitors to peripheral blood.
Aiuti A
The Journal of Experimental Medicine, 185, 111-120 (1997)
A highly efficacious lymphocyte chemoattractant, stromal cell-derived factor 1 (SDF-1)
Bleul CC
The Journal of Experimental Medicine, 184, 1101-1109 (1996)
Stromal cell-derived factor 1a (SDF-1a): A marker of disease burden in patients with atrial fibrillation.
Li D
Scandinavian Cardiovascular Journal : SCJ, 50, 36-41 (2016)
Angiogenic and restorative abilities of human mesenchymal stem cells were reduced following treatment with serum from diabetes mellitus type 2 patients
Rezaie J, et al.
Journal of Cellular Biochemistry, 119(1), 524-535 (2018)
Gene-environment interaction effects on the development of immune responses in the 1st year of life.
Hoffjan S
American Journal of Human Genetics, 76, 696-704 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service