Skip to Content
Merck
All Photos(1)

Key Documents

GF345

Sigma-Aldrich

SDF-1β (CXCL12),
Human Recombinant Animal Free

Synonym(s):

Stromal Cell-Derived Factor, CXCL12, SDF1B, TLSF-b, TPAR1, SDF-1b, TLSF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Level

UniProt accession no.

shipped in

dry ice

General description

SDF-1α and β are stromal derived CXC chemokines, and signal through the CXCR4 receptor. SDF-1α and β chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV suppressive activity in cells expressing the CXCR4 receptor.
Recombinant human SDF-1β is an 8.5 kDa protein containing 72 amino acid residues.

Quality

Determined by its ability to chemoattract human peripheral T cells activated with PHA and IL-2 using a concentration range of 20.0- 80.0 ng/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service