Skip to Content
Merck
All Photos(1)

Key Documents

SRP3140

Sigma-Aldrich

PECAM-I human

recombinant, expressed in HEK 293 cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture

Synonym(s):

CD31 antigen, EndoCAM, Platelet endothelial cell adhesion molecule

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥97% (HPLC)
≥97% (SDS-PAGE)

form

lyophilized

potency

1.0-1.5 μg/mL ED50

mol wt

80.0-95.0 kDa

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PECAM1(5175)

General description

PECAM1 (platelet and endothelial cell adhesion molecule 1), a member of the Ig (immunoglobulin) superfamily, is a multifunctional vascular cell adhesion and signaling molecule. It is a surface molecule found on circulating leukocytes, endothelial cells, and platelets. It is a glycoprotein with a molecular weight of 130kDa, and is composed of a single-chain of six Ig-like extracellular homology domains (total 574-residues), a transmembrane portion of 19 residues, and a cytoplasmic tail containing 118 amino acids. This gene is localized to human chromosome 17q23, and is a 75kb gene composed of 16 exons.
Monomeric glycosylated PECAM-1 migrates at an apparent molecular weight of approximately 80.0-95.0kDa by SDS-PAGE analysis under reducing conditions.

Biochem/physiol Actions

PECAM1 (platelet and endothelial cell adhesion molecule 1) forms an essential part of the endothelial cell intercellular junctions in vascular beds. Its extracellular domain is involved in homo- and heterophilic interactions, and its cytosolic domain is involved in the transduction of a variety of cellular signaling. It controls vascular inflammatory response as it functions as both pro-inflammatory and anti-inflammatory molecule. Its capacity for hemophilic interactions renders it essential for leukocyte transmigration, interendothelial cell bindings, and vascular permeability regulation. This molecule is thought to be involved in the pathogenesis of various disorders such as thrombosis, cardiovascular disease, inflammation, and cancer.

Sequence

ENSFTINSVD MKSLPDWTVQ NGKNLTLQCF ADVSTTSHVK PQHQMLFYKD DVLFYNISSM KSTESYFIPE VRIYDSGTYK CTVIVNNKEK TTAEYQLLVE GVPSPRVTLD KKEAIQGGIV RVNCSVPEEK APIHFTIEKL ELNEKMVKLK REKNSRDQNF VILEFPVEEQ DRVLSFRCQA RIISGIHMQT SESTKSELVT VTESFSTPKF HISPTGMIME GAQLHIKCTI QVTHLAQEFP EIIIQKDKAI VAHNRHGNKA VYSVMAMVEH SGNYTCKVES SRISKVSSIV VNITELFSKP ELESSFTHLD QGERLNLSCS IPGAPPANFT IQKEDTIVSQ TQDFTKIASK SDSGTYICTA GIDKVVKKSN TVQIVVCEML SQPRISYDAQ FEVIKGQTIE VRCESISGTL PISYQLLKTS KVLENSTKNS NDPAVFKDNP TEDVEYQCVA DNCHSHAKML SEVLRVKVIA PVDEVQISIL SSKVVESGED IVLQCAVNEG SGPITYKFYR EKEGKPFYQM TSNATQAFWT KQKASKEQEG EYYCTAFNRA NHASSVPRSK ILTVRVILAP WK

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PECAM-1 Leu125Val (rs688) Polymorphism and Diabetic Nephropathy in Caucasians with Type 2 Diabetes Mellitus.
Zavrsnik M et al
Analytical Cellular Pathology (Amsterdam), 2016, 3152967-3152967 (2016)
Endothelial functions of platelet/endothelial cell adhesion molecule-1 (CD31).
Lertkiatmongkol P et al
Current Opinion in Hematology, 23(3), 253-259 (2016)
Abigail Woodfin et al.
Arteriosclerosis, thrombosis, and vascular biology, 27(12), 2514-2523 (2007-09-18)
Platelet endothelial cell adhesion molecule-1 (PECAM-1 or CD31) is a molecule expressed on all cells within the vascular compartment, being expressed to different degrees on most leukocyte sub-types, platelets, and on endothelial cells where its expression is largely concentrated at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service