Skip to Content
Merck
All Photos(7)

Key Documents

SAB2701819

Sigma-Aldrich

Anti-PCNA antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

cb16, fb36g03, pcna, wu:fa28e03

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

zebrafish, (Japanese Medaka)

technique(s)

immunohistochemistry: suitable
western blot: 500-3000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

zebrafish ... pcna(30678)

General description

Proliferating cell nuclear antigen (PCNA) is a ring-shaped homotrimer protein, that is located at the center of the faithful duplication of eukaryotic genomes. Since it encircles the DNA, PCNA is also known as a sliding clamp. This gene is mapped to human chromosome 20p12.3.

Immunogen

Recombinant protein containing a sequence corresponding to a region within amino acids 31 and 222 of pcna according to NP_571479

Application

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000, WB: 1:500-1:3000, IHC-Wm: 1:50-1:500, Immunostaining: Assay-dependent dilution. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Anti-PCNA antibody has been used in the detection of proliferating cells by immunofluorescence.

Biochem/physiol Actions

This protein is an auxiliary protein of DNA polymerase delta and is involved in the control of eukaryotic DNA replication by increasing the polymerase′s processibility during elongation of the leading strand.
Proliferating cell nuclear antigen (PCNA) controls the production of the leading and lagging strands, which are required for the duplication of DNA. It acts as a cell cycle regulatory protein. This protein regulates apoptosis. PCNA is also involved in non-replicative DNA synthesis events.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 1% BSA, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Forging ahead through darkness: PCNA, still the principal conductor at the replication fork.
Katherine NC and George-Lucian M
Molecular Cell, 65(3), 380?392-380?392 (2017)
Fate bias during neural regeneration adjusts dynamically without recapitulating developmental fate progression.
Kei JNC, et al.
Neural Dev., 12(1), 12-12 (2017)
PCNA (proliferating cell nuclear antigen)
Stoimenov I and Helleday T
Atlas of Genetics and Cytogenetics in Oncology and Haematology, 16(3) (2012)
Luca Di Cesare et al.
Journal of morphology, 284(9), e21630-e21630 (2023-08-16)
In viviparous teleosts, intraovarian gestation occurs intrafollicularly, as in poeciliids, or intraluminally, as in goodeids and anablepids. Furthermore, there are two different forms of embryonic nutrition: lecithotrophy and matrotrophy; depending on the species, these can be exclusive or coexist during
Francisco Acuña et al.
Journal of morphology, 281(7), 710-724 (2020-05-08)
The uterus is an organ with great plasticity due to the morphological and physiological changes it experiences during the estrous cycle and pregnancy. In mammals, pregnancy requires diverse sex hormones, growth factors and cytokines, among others, for promoting uterine remodeling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service