Skip to Content
Merck
All Photos(1)

Key Documents

SRP3080

Sigma-Aldrich

IL-17 human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CTLA-8, IL-17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

31.0 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

off-white to yellow

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL17A(3605)

General description

The gene IL17A (interleukin 17A), also referred to as IL-17, is mapped to human chromosome 6p12.2. The gene contain three exons and encodes a proinflammatory cytokine produced by activated T cells.

Biochem/physiol Actions

The cytokine interleukin 17A is involved in the activation of proinflammatory cytokines, chemokines, and cell adhesion molecules in several cell types such as, macrophages, dendritic cells, T cells, synovial cells, and endothelial cells. Overexpression of this protein has been associated with rheumatoid arthritis, systemic lupus erythematosus, Behcet′s disease, allograft rejection, nephritic syndrome, asthma, and multiple sclerosis. In mice, it has been observed to be implicated in the development of allergic and autoimmune diseases. Polymorphism in IL17A gene has been associated with lung cancer.

Sequence

MIVKAGITIP RNPGCPNSED KNFPRTVMVN LNIHNRNTNT NPKRSSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CINADGNVDY HMNSVPIQQE ILVLRREPPH CPNSFRLEKI LVSVGCTCVT PIVHHVA

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin-17A and -17F genes polymorphisms in lung cancer.
Kaabachi W
Cytokine, 66, 23-29 (2014)
IL-17 plays an important role in the development of experimental autoimmune encephalomyelitis.
Komiyama Y
Journal of Immunology, 177, 566-573 (2006)
Hye-Lin Ha et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(33), E3422-E3431 (2014-08-06)
Psoriasis is a chronic inflammatory skin disease characterized by abnormal keratinocyte proliferation and differentiation and by an influx of inflammatory cells. The mechanisms underlying psoriasis in humans and in mouse models are poorly understood, although evidence strongly points to crucial
Junhua Wang et al.
PLoS neglected tropical diseases, 9(5), e0003755-e0003755 (2015-05-09)
The growth potential of the tumor-like Echinococcus multilocularis metacestode (causing alveolar echinococcosis, AE) is directly linked to the nature/function of the periparasitic host immune-mediated processes. We previously showed that Fibrinogen-like-protein 2 (FGL2), a novel CD4+CD25+ Treg effector molecule, was over-expressed
Wei Peng et al.
Frontiers in oncology, 12, 781093-781093 (2022-04-12)
This study aimed to investigate the effect of OCT4&SOX2 specific cytotoxic T lymphocytes (CTLs) plus programmed cell death protein-1 (PD-1) inhibitor (nivolumab) on treating breast cancer stem-like cells (BCSCs) in vitro and drug-resistance breast cancer (DRBC) mice in vivo. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service