Skip to Content
Merck
All Photos(2)

Key Documents

SML3666

Sigma-Aldrich

AMP-Deoxynojirimycin

≥97% (HPLC)

Synonym(s):

AMP-dNJ, AMP-dNM, (2R,3R,4R,5S)-2-(Hydroxymethyl)-1-[5-(tricyclo[3.3.1.13,7]dec-1-ylmethoxy)pentyl]-3,4,5-piperidinetriol, AMP-DNJ, AMP-DNM, Adamantane-pentyl-DNM, Adamantane-pentyl-dNM, MZ-21, N-(5′-Adamantane-1′-yl-methoxy)pentyl-1-deoxynojirimycin; (2R,3R,4R,5S)-1-[5-(1-Adamantylmethoxy)pentyl]-2-(hydroxymethyl)piperidine-3,4,5-triol, N-(5-Adamantane-1-yl-methoxy)pentyl-1-deoxynojirimycin; N-(5-Adamantane-1-yl-methoxy)pentyl)-DNM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H39NO5
CAS Number:
Molecular Weight:
397.55
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O[C@@H]1[C@H](N(C[C@@H]([C@H]1O)O)CCCCCOCC2(C3)C[C@@H](C4)C[C@H]3C[C@@H]4C2)CO

InChI

1S/C22H39NO5/c24-13-18-20(26)21(27)19(25)12-23(18)4-2-1-3-5-28-14-22-9-15-6-16(10-22)8-17(7-15)11-22/h15-21,24-27H,1-14H2/t15-,16+,17-,18-,19+,20-,21-,22?/m1/s1

InChI key

XVYLNHVEAOOEGI-OIPNBASOSA-N

Biochem/physiol Actions

AMP-deoxynojirimycin (AMP-DNM) is an orally available iminosugar that acts as a reversible, potent and selective inhibitor against the non-lysosomal glucosylceramidase GBA2 (IC50 = 1.7 nM) over the glucosylceramide synthase (GCS, UGCG), ER alpha-glucosidases, the lysosomal glucocerebrosidase GBA (IC50 = 0.16 μM) and alpha-glucosidase GAA (IC50 = 0.87 μM). AMP-DNM selectively inhibits cellular GBA2 in cultures (GBA2/GBA IC50 = 0.3 nM/100 nM; human melanoma cells) and exhibits in vivo therapeutic efficacy in murine models of obesity (100 mg/kg/day p.o.) and colitis (10-50 mg/kg/day via i.p. or 1.25-6.25g/kg chow).

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of glycolipid biosynthesis by N-(5-adamantane-1-yl-methoxy-pentyl)-deoxynojirimycin protects against the inflammatory response in hapten-induced colitis
International Immunopharmacology, 4(7), 939-951 (2004)
Reduction of glycosphingolipid biosynthesis stimulates biliary lipid secretion in mice
Hepatology, 49(2), 637-645 (2009)
Cytosolic glucosylceramide regulates endolysosomal function in Niemann-Pick type C disease
Neurobiology of Disease, 127, 242-252 (2019)
Species-specific differences in nonlysosomal glucosylceramidase GBA2 function underlie locomotor dysfunction arising from loss-of-function mutations
The Journal of Biological Chemistry, 294(11), 3853-3871 (2019)
Generation of specific deoxynojirimycin-type inhibitors of the non-lysosomal glucosylceramidase
The Journal of biological chemistry, 273(41), 26522-26527 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service