Skip to Content
Merck
All Photos(1)

Key Documents

SML3522

Sigma-Aldrich

DT2216

≥90% (HPLC)

Synonym(s):

(2S,4R)-1-((S)-2-(7-(4-((R)-3-((4-(N-(4-(4-((4′-chloro-4,4-dimethyl-3,4,5,6-tetrahydro-[1,1′-biphenyl]-2-yl)methyl)piperazin-1-yl)benzoyl)sulfamoyl)-2-((trifluoromethyl)sulfonyl)phenyl)amino)-4-(phenylthio)butyl)piperazin-1-yl)-7-oxoheptanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide, DT 2216, DT-2216

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C77H96ClF3N10O10S4
CAS Number:
Molecular Weight:
1542.36
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

CC(C)(C1)CCC(C2=CC=C(C=C2)Cl)=C1CN3CCN(C4=CC=C(C=C4)C(NS(=O)(C5=CC(S(C(F)(F)F)(=O)=O)=C(C=C5)N[C@@H](CSC6=CC=CC=C6)CCN7CCN(CC7)C(CCCCCC(N[C@@H](C(C)(C)C)C(N8[C@@H](C[C@H](C8)O)C(N[C@H](C9=CC=C(C%10=C(N=CS%10)C)C=C9)C)=O)=O)=O)=O)=O)=O)CC3

InChI

1S/C77H96ClF3N10O10S4/c1-51(53-18-20-55(21-19-53)70-52(2)82-50-103-70)83-73(96)66-44-61(92)48-91(66)74(97)71(75(3,4)5)85-68(93)16-12-9-13-17-69(94)90-42-36-87(37-43-90)35-33-59(49-102-62-14-10-8-11-15-62)84-65-31-30-63(45-67(65)104(98,99)77(79,80)81)105(1

InChI key

PXVFFBGSTYQHRO-REQIQPEASA-N

Biochem/physiol Actions

DT2216 is a selective BCL-XL, but not BCL-2 or BCL-W, degrader composed of a von Hippel-Lindau (VHL) E3 ligase-targeting ligand linked to a ABT-263 (navitoclax) derivative. DT2216 is more potent than ABT-263 against BCL-XL-dependent leukemia (MOLT-4 IC50 = 52 vs 191 nM) and cancer cells (MDA-MB-231 IC50 = 229 vs 707 nM). DT2216 effectively inhibits the growth of several xenograft tumors in vivo either alone (15 mg/kg/wk i.p.) or in combination with other chemotherapeutic agents (docetaxel or VDL), exhbiting improved efficacy and reduced thrombocytopenia when compared with ABT-263 as a result of greatly reduced platelet toxicity due to poor VHL expression in platelets.
Selective BCL-XL degrader composed of a von Hippel-Landau (VHL) E3 ligase-targeting ligand linked to a ABT-263 (navitoclax) derivative.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overcoming Gemcitabine Resistance in Pancreatic Cancer Using the BCL-XL-Specific Degrader DT2216
Dinesh Thummuri, Sajid Khan, Janet Wiegand
Molecular Cancer Therapeutics, 21, 184-192 (2022)
BCL-XL PROTAC degrader DT2216 synergizes with sotorasib in preclinical models of KRASG12C-mutated cancers
Sajid Khan
Journal of Hematology & Oncology, 15, 23-23 (2022)
Ryan Kolb et al.
Nature communications, 12(1), 1281-1281 (2021-02-26)
Regulatory T cells (Tregs) play an important role in maintaining immune homeostasis and, within tumors, their upregulation is common and promotes an immunosuppressive microenvironment. Therapeutic strategies that can eliminate Tregs in the tumor (i.e., therapies that do not run the
Yonghan He et al.
Journal of hematology & oncology, 13(1), 95-95 (2020-07-18)
Patients with advanced T cell lymphomas (TCLs) have limited therapeutic options and poor outcomes in part because their TCLs evade apoptosis through upregulation of anti-apoptotic Bcl-2 proteins. Subsets of TCL cell lines, patient-derived xenografts (PDXs), and primary patient samples depend
Sajid Khan et al.
Nature medicine, 25(12), 1938-1947 (2019-12-04)
B-cell lymphoma extra large (BCL-XL) is a well-validated cancer target. However, the on-target and dose-limiting thrombocytopenia limits the use of BCL-XL inhibitors, such as ABT263, as safe and effective anticancer agents. To reduce the toxicity of ABT263, we converted it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service