Skip to Content
Merck
All Photos(1)

Key Documents

SML2938

Sigma-Aldrich

Vernakalant hydrochloride

≥98% (HPLC)

Synonym(s):

(3R)-1-((1R,2R)-2-(2-(3,4-Dimethoxyphenyl)ethoxy)cyclohexyl)pyrrolidin-3-ol hydrochloride, (3R)-1-[(1R,2R)-2-[2-(3,4-Dimethoxyphenyl)ethoxy]cyclohexyl]-3-pyrrolidinol hydrochloride, RSD 1235, RSD-1235, RSD1235

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31NO4 · HCl
CAS Number:
Molecular Weight:
385.93
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

Cl.N3(C[C@@H](CC3)O)[C@H]1[C@@H](CCCC1)OCCc2cc(c(cc2)OC)OC

InChI

1S/C20H31NO4.ClH/c1-23-19-8-7-15(13-20(19)24-2)10-12-25-18-6-4-3-5-17(18)21-11-9-16(22)14-21;/h7-8,13,16-18,22H,3-6,9-12,14H2,1-2H3;1H/t16-,17-,18-;/m1./s1

InChI key

JMHYCBFEEFHTMK-IIUXMCBISA-N

Biochem/physiol Actions

Anti-arrhythmic that exerts anti-fibrillatory efficacy via potential- & rate-dependent Nav1.5 (SCN5A) as well as atrial-selective Kv1.5 (KCNA5) blockade in vivo.
Vernakalant is a multiple ion channel blocker that exerts in vivo anti-fibrillatory (anti-arrhythmic) efficacy (ED50 = 1.5 μmol/kg/min iv. against ischemia-induced arrhythmias in rats) via atrial-selective Kv1.5 blockage (hKv1.5/rKv4.2/rKv4.3 IC50 = 13/38/30 μM at 1Hz & -80 to 60 mV in 200 (Kv1.5) or 400 (Kv4) msec) as well as potential- and rate-dependent Nav1.5 blockade (inward Na current INa IC50 = 9 μM/20 Hz & -80 mV, 31 μM/1Hz & -60 mV, 107 μM/1Hz & -120 mV using HEK hNav1.5 cells).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonas Goldin Diness et al.
Circulation. Arrhythmia and electrophysiology, 10(10) (2017-10-12)
Evidence has emerged that small-conductance Ca2+-activated K+ (SK) channels constitute a new target for treatment of atrial fibrillation (AF). SK channels are predominantly expressed in the atria as compared with the ventricles. Various marketed antiarrhythmic drugs are limited by ventricular
Gerrit Frommeyer et al.
Journal of cardiovascular electrophysiology, 27(10), 1214-1219 (2016-06-11)
Ranolazine has been reported to have an antiarrhythmic potential. The aim of this study was to assess the electrophysiologic effects of ranolazine and to compare its effects to vernakalant in an experimental whole-heart model of short-QT syndrome. Rabbit hearts were
Gerrit Frommeyer et al.
Europace : European pacing, arrhythmias, and cardiac electrophysiology : journal of the working groups on cardiac pacing, arrhythmias, and cardiac cellular electrophysiology of the European Society of Cardiology, 19(5), 866-873 (2016-10-06)
The antiarrhythmic drug vernakalant exerts antiarrhythmic effects in atrial fibrillation. Recent experimental data suggest interactions with the late sodium current and antiarrhythmic effects in ventricular arrhythmias. We aimed at investigating whether treatment with vernakalant reduces polymorphic ventricular tachycardia (VT) in
Rong Chen et al.
Biochemistry, 57(18), 2704-2710 (2018-04-14)
Molecular dynamics simulations are employed to determine the inhibitory mechanisms of three drugs, 5-(4-phenoxybutoxy)psoralen (PAP-1), vernakalant, and flecainide, on the voltage-gated K+ channel Kv1.5, a target for the treatment of cardiac arrhythmia. At neutral pH, PAP-1 is neutral, whereas the
Toshiki Chiba et al.
Journal of pharmacological sciences, 130(3), 170-176 (2016-04-02)
Electrical remodeling plays a pivotal role in maintaining the reentry during atrial fibrillation. In this study, we assessed influence of electrical remodeling on pharmacological manipulation of the atrial refractoriness in rabbits. We used an atrial electrical remodeling model of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service