Skip to Content
Merck
All Photos(1)

Key Documents

SML2485

Sigma-Aldrich

Marbofloxacin

≥98% (HPLC)

Synonym(s):

9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[3,2,1-ij][4,1,2]benzoxadiazine-6-carboxylic acid, Aurizon, Marbocyl, Zeniquin, 9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-piperazino)-7-oxo-7H-pyrido[1,2,3-ij][1,2,4]benzoxadiazine-6-carboxylic acid, Zeniquin®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19FN4O4
CAS Number:
Molecular Weight:
362.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN1CCN(CC1)c2c(F)cc3C(=O)C(=CN4N(C)COc2c34)C(O)=O

InChI

1S/C17H19FN4O4/c1-19-3-5-21(6-4-19)14-12(18)7-10-13-16(14)26-9-20(2)22(13)8-11(15(10)23)17(24)25/h7-8H,3-6,9H2,1-2H3,(H,24,25)

InChI key

BPFYOAJNDMUVBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Marbofloxacin is a broad-spectrum fluoroquinolone antibiotic active against both Gram-negative and Gram-positive bacteria. It is believed to act by impairing the bacterial DNA gyrase. Marbofloxacin has also been shown to exhibit antileishmanial activity in a canine model.

Legal Information

Zeniquin is a registered trademark of Pfizer, Inc.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Spreng et al.
Journal of veterinary pharmacology and therapeutics, 18(4), 284-289 (1995-08-01)
Marbofloxacin is a new fluoroquinolone developed exclusively for veterinary use. Minimum inhibitory concentrations of marbofloxacin were assessed for 816 recent isolates associated with canine or feline diseases. Marbofloxacin showed a broad spectrum of activity against gram-negative and gram-positive bacteria. In
Marilisa Novacco et al.
Veterinary microbiology, 217, 112-120 (2018-04-05)
Mycoplasma haemofelis is the most pathogenic feline hemoplasma species and a causative agent of infectious hemolytic anemia in cats. Current treatment protocols are effective in reducing M. haemofelis blood loads and clinical signs but consistent bacteremia clearance is rarely achieved.
Carmen Pineda et al.
PloS one, 12(10), e0185981-e0185981 (2017-10-06)
Treatment of canine leishmaniasis (CanL) represents a challenge. Due to the high prevalence of renal disease associated to CanL, it is important to find an effective drug that does not damage the kidneys. Marbofloxacin has been shown to be effective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service