Skip to Content
Merck
All Photos(1)

Documents

SML2107

Sigma-Aldrich

Tulathromycin A

≥95% (HPLC), powder, macrolide antibiotic

Synonym(s):

(2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-13-[[2,6-dideoxy-3-C-methyl-3-O-methyl-4-C-[(propylamino)methyl]-a-L-ribo-hexopyranosyl]oxy]-2-ethyl-3,4,10-trihydroxy-3,5,8,10,12,14-hexamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-b-D-xylo-hexopyranosyl]oxy]-1-oxa-6-azacyclopentadecan-15-one, CP-472295

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H79N3O12
CAS Number:
Molecular Weight:
806.08
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Tulathromycin A, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@@]([C@H](OC([C@@H]([C@H]1O[C@]2([H])O[C@@H](C)[C@@](O)(CNCCC)[C@@](C2)(OC)C)C)=O)CC)([C@@H]([C@H](NC[C@H](C)C[C@]([C@@H]([C@H]1C)O[C@]3([H])O[C@H](C)C[C@H](N(C)C)[C@H]3O)(O)C)C)O)O

InChI

1S/C41H79N3O12/c1-15-17-42-22-41(50)28(8)53-31(20-39(41,10)51-14)55-33-25(5)35(56-37-32(45)29(44(12)13)18-24(4)52-37)38(9,48)19-23(3)21-43-27(7)34(46)40(11,49)30(16-2)54-36(47)26(33)6/h23-35,37,42-43,45-46,48-50H,15-22H2,1-14H3/t23-,24-,25+,26-,27-,28+,29+,30-,31+,32-,33+,34-,35-,37+,38-,39-,40-,41+/m1/s1

InChI key

GUARTUJKFNAVIK-QPTWMBCESA-N

Biochem/physiol Actions

Tulathromycin A is a macrolide antibiotic. It binds to the subunit 50S of the bacterial ribosome, interfering with bacterial protein synthesis.
Tulathromycin is a triamilide antimicrobial, which is structurally similar to the di-basic azalide antibiotic azithromycin. It is used to treat bovine and swine bacterial respiratory diseases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nigel A Evans
Veterinary therapeutics : research in applied veterinary medicine, 6(2), 83-95 (2005-08-12)
Tulathromycin is a novel triamilide antimicrobial that has been approved for use in the treatment and prevention of bovine respiratory disease and the treatment of swine respiratory disease in the European Union and the United States. The agent penetrates gram-negative
Analytical procedure for the determination of tulathromycin in swine plasma
Gajda A, et al.
Bulletin of the Veterinary Institute in Pulawy, 57(2), 191-195 (2013)
Diana Gáler et al.
Journal of agricultural and food chemistry, 52(8), 2179-2191 (2004-04-15)
Tulathromycin is a novel member of the triamilide class of antibiotics that was developed as a safe and effective single-dose treatment of bovine and porcine respiratory disease. An accurate and precise analytical method was developed for the extraction and measurement
Tulathromycin: An overview of a new triamilide antimicrobial for livestock respiratory disease
Evans NA, et al.
Veterinary Therapeutics : Research in Applied Veterinary Medicine, 6(2), 83-83 (2005)
Haihong Hao et al.
Frontiers in microbiology, 7, 477-477 (2016-04-20)
To evaluate microbiological safety of tulathromycin on human intestinal bacteria, tulathromycin (0, 0.1, 1, 10, and 100 μg/mL) was added into Chemostats. Before and after drug exposure, we monitored (1) population, SCFA products, antimicrobial resistance, and colonization resistance of gut

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service