Skip to Content
Merck
All Photos(1)

Key Documents

SML2355

Sigma-Aldrich

BIX02189

≥98% (HPLC)

Synonym(s):

(3Z)-3-[[[3-[(Dimethylamino)methyl]phenyl]amino]phenylmethylene]-2,3-dihydro-N,N-dimethyl-2-oxo-1H-indole-6-carboxamide, (Z)-3-((3-((Dimethylamino)methyl)phenylamino)(phenyl)methylene)-N,N-dimethyl-2-oxoindoline-6-carboxamide, BIX 02189, BIX-02189

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28N4O2
CAS Number:
Molecular Weight:
440.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CN(C)CC1=CC=CC(N/C(C2=CC=CC=C2)=C(C3=CC=C(C(N(C)C)=O)C=C3N4)\C4=O)=C1

InChI

1S/C27H28N4O2/c1-30(2)17-18-9-8-12-21(15-18)28-25(19-10-6-5-7-11-19)24-22-14-13-20(27(33)31(3)4)16-23(22)29-26(24)32/h5-16,28H,17H2,1-4H3,(H,29,32)/b25-24-

InChI key

HOMJAAIVTDVQJA-IZHYLOQSSA-N

Biochem/physiol Actions

BIX02189 is an ATP site-targeting, potent and selective MEK5 inhibitor (IC50 =1.5 nM, [ATP] = 750 nM) with much reduced or no potency against 79 other kinases (CSF1R (FMS)/ERK5/Lck/Jak3/TGFβR1/RPS6KA6 (RSK4) IC50 = 46/59/250/440/580/990 nM, MEK1/MEK2/ERK2/JNK2/EGFR/STK16 IC50 >6.2 μM). BIX02189 blocks cellular ERK5, but not ERK1/2 or p38, phosphorylation induction (IC50 <1 μM in sorbitol-stimulated HeLa), as well as downstream MEF2C transcriptional activation (IC50 = 0.53/0.26 μM, MEK5/ERK5/MEF2C HeLa/HEK293 reporter cells). BIX02189 in vivo efficacies are demonstrated in murine models of atherosclerosis, cancer, and diabetes (10-20 mg/kg i.p.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seong Ji Park et al.
Cancer letters, 381(2), 314-322 (2016-08-21)
Transforming growth factor-β1 (TGF-β1) promotes tumor metastasis by inducing an epithelial-to-mesenchymal transition (EMT) in cancer cells. In this study, we investigated the effects of BIX02189 and XMD8-92, pharmacologic inhibitors of the MEK5 [mitogen-activated protein kinase/extracellular-signal-regulated kinase (ERK)5] signaling pathway, on
Qi Zhao et al.
Oncotarget, 9(1), 1229-1236 (2018-02-09)
Diabetic retinopathy (DR) is a major complication of diabetes, and causes pathological changes in retina blood vessels, as the most common cause of vision loss. Extracellular-signal-regulated kinase 5 (ERK5) is the newest discovered member in the mitogen-activated protein kinases (MAPKs)
Carolyn J Loveridge et al.
Scientific reports, 7(1), 13241-13241 (2017-10-19)
Extracellular signal-regulated protein kinase 5 (ERK5) has been implicated during development and carcinogenesis. Nkx3.1-mediated Cre expression is a useful strategy to genetically manipulate the mouse prostate. While grossly normal at birth, we observed an unexpected phenotype of spinal protrusion in
Dae-Hwan Nam et al.
Molecules and cells, 40(7), 457-465 (2017-07-07)
Streptozotocin (STZ)-induced murine models of type 1 diabetes have been used to examine ER stress during pancreatic β-cell apoptosis, as this ER stress plays important roles in the pathogenesis and development of the disease. However, the mechanisms linking type 1
Congde Chen et al.
Cell proliferation, 51(3), e12410-e12410 (2017-11-22)
Restoring a functional beta-cell mass is a fundamental goal in treating diabetes. A complex signalling pathway network coordinates the regulation of beta-cell proliferation, although a role for ERK5 in this network has not been reported. This question was addressed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service