Skip to Content
Merck
All Photos(1)

Documents

SML2049

Sigma-Aldrich

AR-R17779 Hydrochloride

≥97% (HPLC)

Synonym(s):

(-)-AR-R 13489 HCl, (-)-AR-R13489 HCl, (-)-Spiro[1-azabicyclo[2.2. 2]octane-3,5′-oxazolidin]-2′-one hydrochloride; (3S)-Spiro[1-azabicyclo[2.2.2]octane-3,5′-oxazolidine]-2′-one hydrochloride, (S)-AR-R 13489 HCl, (S)-AR-R13489 HCl, AR-R 17779 HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N2O2 · HCl
CAS Number:
Molecular Weight:
218.68
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

optical activity

[α]/D -59 to -69°, c = 1.0 in methanol

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1O[C@@]2(CN1)CN3CCC2CC3.Cl

InChI

1S/C9H14N2O2.ClH/c12-8-10-5-9(13-8)6-11-3-1-7(9)2-4-11;/h7H,1-6H2,(H,10,12);1H/t9-;/m0./s1

InChI key

XGLBLUBBDSJBIU-FVGYRXGTSA-N

Biochem/physiol Actions

AR-R17779 is a nicotinic acetylcholine receptor alpha7 full agonist that targets α7 nAChR with high affinity (Ki = 92 nM/rat α7 against 5 nM α-BTX vs.16 μM/rat α4β2 against 3 nM (-)-nicotine) and selectivity (EC50 = 6.2/10/12.7 μM using human/rat/monkey α7 nAChR-Xenopus oocyte by whole cell voltage clamp, no antagonistic activity against acetylcholine using human α4β2-, α3β4-, α3β2-, α3β2α5-expressing oocytes or antagonistic activity against 5-HT using rat 5HT3a-exxpressing oocytes). AR-R17779 exhibits cognition-improving efficacy in rats (1-20 mg/kg s.c) and mice (1-20 mg/kg i.p.) in vivo and is widely employed for studying other α7 nAChR-dependent physiological functions.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F A Koopman et al.
Oral diseases, 21(7), 858-865 (2015-06-23)
Activation of the cholinergic anti-inflammatory pathway (CAP) has been shown to reduce inflammation in animal models, while abrogation of the pathway increases inflammation. We investigated whether modulation of CAP influences inflammation in the non-obese diabetic (NOD) mouse model for Sjögren's
Roger L Papke et al.
European journal of pharmacology, 524(1-3), 11-18 (2005-11-04)
An alpha7 nicotinic acetylcholine receptor sequence was cloned from Rhesus monkey (Macaca mulatta). This clone differs from the mature human alpha7 nicotinic acetylcholine receptor in only four amino acids, two of which are in the extracellular domain. The monkey alpha7
E D Levin et al.
Behavioural pharmacology, 10(6-7), 675-680 (2000-04-26)
Nicotinic acetylcholine systems have been found to be important for learning and memory function. The prototypic nicotinic agonist nicotine has been shown in a variety of studies to improve aspects of cognitive function. The specific involvement of nicotinic receptor subtypes
Lei Zhao et al.
Toxicological sciences : an official journal of the Society of Toxicology, 153(1), 103-111 (2016-06-23)
Maternal cigarette smoke is the major risk of sudden infant death syndrome (SIDS). A depressed ventilatory response to hypoxia (HVR) and hypercapnia (HCVR) is thought to be responsible for the pathogenesis of SIDS and the carotid body is critically involved
Aya Watanabe et al.
Atherosclerosis, 244, 113-120 (2015-11-28)
Activation of vagal nerve suppresses inflammatory responses through activation of α7 nicotinic acetylcholine receptor (nAchR). We sought to determine whether AR-R17779, a selective agonist of α7nAchR, affects the development of abdominal aortic aneurysm (AAA). AAA was induced by topical application

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service