Skip to Content
Merck
All Photos(1)

Key Documents

SML1695

Sigma-Aldrich

RSV604

≥98% (HPLC)

Synonym(s):

(S)-1-(2-Fluorophenyl)-3-(2-oxo-5-phenyl-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)-urea, RSV-604

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17FN4O2
CAS Number:
Molecular Weight:
388.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

film

storage condition

protect from light

color

white

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1)[C@@H](NC(NC2=CC=CC=C2F)=O)N=C(C3=CC=CC=C3)C4=C1C=CC=C4

InChI key

MTPVBMVUENFFLL-HXUWFJFHSA-N

Biochem/physiol Actions

RSV604 (RSV-604) is a cell-permeable, non-cytotoxic (CC50 >50 μM) benzodiazepine derivative that is reported to target respiratory syncytial virus (RSV) nucleoprotein (N) via affinity interaction (Kd = 134 μM) and display similar antiviral potency against 40 clinical human RSV isolates (Ave. EC50 = 800 nM), including both subtypes A and B. Although RSV604 displays host-dependent potency against RSV cellular replication (EC50 = 2 μM/HeLa, 1.8 μM/HEp-2, >50 μM/BHK-21 based on cellular viral F protein level 72 hrs post RSV A2 infection) and release (by >1,000,000-fold/HeLa vs. no effect/BHK-21 72 hrs post infection; [RSV604] = EC90), complete loss of infectivity of released RSV from RSV604-treated cells is observed in both HeLa and BHK-21 cultures. RSV604 is also active against bovine RSV, but inactive toward mouse pneumonia virus, human metapneumovirus or parainfluenza virus types 1 &3 (PIV1 &PIV3).
RSV604 (RSV-604) is a cell-permeable, non-cytotoxic benzodiazepine derivative that is reported to target respiratory syncytial virus (RSV) nucleoprotein (N) via affinity interaction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuzhen Gao et al.
Molecules (Basel, Switzerland), 26(9) (2021-05-06)
Respiratory syncytial virus (RSV) is a major pathogen that causes severe lower respiratory tract infection in infants, the elderly and the immunocompromised worldwide. At present no approved specific drugs or vaccines are available to treat this pathogen. Recently, several promising

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service