MAB5504
Anti-Vesicular Glutamate Transporter 2 Antibody
clone 8G9.2, Chemicon®, from mouse
Synonym(s):
VGLUT2
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
8G9.2, monoclonal
species reactivity
mouse, rat
species reactivity (predicted by homology)
human
manufacturer/tradename
Chemicon®
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... SLC17A6(57084)
General description
Vesicular Glutamate Transporter 2 (VGLUT2) is a sodium dependent inorganic phosphate cotransporter that is involved in the calcium dependent glutamate release from astrocytes. It is expressed in only a subset of neurons, localized to synaptic vesicles, at synapses exhibiting classical excitatory features. VGLUT2 mRNA is found in brain regions that lack VGLUT1, a related protein also displaying sodium dependent inorganic phosphate cotransporter activity.
Specificity
Reacts with VGLUT2 (Vesicular Glutamate Transporter 2).
Application
Detect Vesicular Glutamate Transporter 2 using this Anti-Vesicular Glutamate Transporter 2 Antibody validated for use in WB, IH.
Immunohistochemistry: A 1:50-1:300 dilution of a previous lot worked using ABC with NiDAB on floating mouse brain sections.
Optimal working dilutions must be determined by end user.
Optimal working dilutions must be determined by end user.
Quality
Evaluated by Western Blot on Mouse brain lysates.
Western Blotting Analysis: 1:500 dilution of this antibody detected VGLUT2 on 10 µg of Mouse brain lysates.
Western Blotting Analysis: 1:500 dilution of this antibody detected VGLUT2 on 10 µg of Mouse brain lysates.
Target description
56 kDa
Physical form
Format: Purified
Mouse monoclonal in PBS containing 0.1% sodium azide.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
recommended
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Purkinje cell ataxin-1 modulates climbing fiber synaptic input in developing and adult mouse cerebellum.
Ebner, BA; Ingram, MA; Barnes, JA; Duvick, LA; Frisch, JL; Clark, HB; Zoghbi, HY; Ebner, TJ; Orr, HT
The Journal of Neuroscience null
I Pose et al.
Neuroscience, 172, 246-255 (2010-11-04)
The rostral ventro-medial medullary reticular formation is a complex structure that is involved with a variety of motor functions. It contains glycinergic neurons that are activated during active (rapid eye movement (REM)) sleep (AS); these neurons appear to be responsible
Peiyan Wong et al.
The Journal of comparative neurology, 510(4), 440-461 (2008-07-31)
The temporal cortex of grey squirrels contains three architectonically distinct regions. One of these regions, the temporal anterior (Ta) region has been identified in previous physiological and anatomical studies as containing several areas that are largely auditory in function. Consistent
SCA1-like disease in mice expressing wild-type ataxin-1 with a serine to aspartic acid replacement at residue 776.
Duvick, L; Barnes, J; Ebner, B; Agrawal, S; Andresen, M; Lim, J; Giesler, GJ; Zoghbi, HY; Orr, HT
Neuron null
Chia-Chi Liao et al.
The Journal of comparative neurology, 524(7), 1494-1526 (2015-11-01)
A complete unilateral lesion of the dorsal column somatosensory pathway in the upper cervical spinal cord deactivates neurons in the hand region in contralateral somatosensory cortex (areas 3b and 1). Over weeks to months of recovery, parts of the hand
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service