Skip to Content
Merck

Key Documents

Skip To

F9397

Flutamide

Non-steroidal anti-androgen drug

Synonym(s):

2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C11H11F3N2O3
CAS Number:
Molecular Weight:
276.21
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
51111800
EC Number:
236-341-9
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Flutamide,

Quality Segment

originator

Schering Plough

SMILES string

CC(C)C(=O)Nc1ccc(c(c1)C(F)(F)F)[N+]([O-])=O

InChI

1S/C11H11F3N2O3/c1-6(2)10(17)15-7-3-4-9(16(18)19)8(5-7)11(12,13)14/h3-6H,1-2H3,(H,15,17)

InChI key

MKXKFYHWDHIYRV-UHFFFAOYSA-N

Gene Information

human ... AR(367)
mouse ... Ar(11835)
rat ... Ar(24208)

Application

Flutamide has been used to effect of testosterone on Glial cell-derived neurotrophic factor (Gdnf) mRNA production in vitro. It has also been used to study the effects of flutamide on anogenital distance and nipple retention.

Biochem/physiol Actions

Flutamide is a non-steroidal anti-androgen drug. It consists of a nitroaromatic structure. Flutamide is a potent competitor of testosterone and dihydrotestosterone receptors. It is a potent hepatotoxin.

Features and Benefits

This compound was developed by Schering Plough. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F02856001285851SML1626
description

-

description

European Pharmacopoeia (EP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

≥98% (HPLC)

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

100

originator

Schering Plough

originator

-

originator

-

originator

-

Gene Information

human ... AR(367)
mouse ... Ar(11835)
rat ... Ar(24208)

Gene Information

human ... AR(367)

Gene Information

human ... AR(367)

Gene Information

-


Still not finding the right product?

Explore all of our products under Flutamide


Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Liver

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

  1. How Can I prepare 1nM solution of flutamide? flutamide dissolves in ethanol? AFTER DISSOLVING flutamide IN ETHANOL , WHATS the storage conditions and useful life?

    1 answer
    1. Flutamide is soluble in ethanol (50 mg/ml) with heat as needed, yielding a clear to hazy, yellow to yellow-green solution. It is also soluble in acetone, ethyl acetate, methanol, chloroform, and ether.

      The molecular weight of flutamide is 276.21, and this value can be used to calculate the amount needed to make a 1nM solution in the desired volume:
      https://www.sigmaaldrich.com/support/calculators-and-apps/mass-molarity-calculator

      Using freshly made solutions whenever possible is always recommended, but storing them at -20°C in non-diluted aliquots should maintain their stability for at least one month.

      Helpful?

Reviews

No rating value

Active Filters