Skip to Content
Merck
All Photos(1)

Key Documents

SML1240

Sigma-Aldrich

Fondaparinux sodium

≥95% (HPLC)

Synonym(s):

Fondaparin sodium, Methyl O-2-deoxy-6-O-sulfo-2-(sulfoamino)-α-D-glucopyranosyl-(14)-O-β-D-glucopyranuronosyl-(14)-O-2-deoxy-3,6-di-O-sulfo-2-(sulfoamino)-α-D-glucopyranosyl-(14)-O-2-O-sulfo-α-L-idopyranuronosyl-(14)-2-deoxy-2-(sulfoamino)-α-D-glucopyranoside 6-(hydrogen sulfate) sodium salt, SR-90107A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H43N3O49S8 · 10Na
CAS Number:
Molecular Weight:
1728.08
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D 42.0 to 55.0° (c = 0.5 in water)

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[S](=O)(=O)(N[C@H]1[C@H](O[C@@H]([C@H]([C@@H]1O[S](=O)(=O)[O-])O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O[C@H]5O[C@@H]([C@H]([C@@H]([C@H]5N[S](=O)(=O)[O-])O)O)CO[S](=O)(=O)[O-])C(=O)[O-])CO[S](=O)(

InChI

1S/C31H53N3O49S8.10Na/c1-69-27-9(33-85(48,49)50)13(37)17(6(74-27)3-71-88(57,58)59)76-31-22(83-91(66,67)68)16(40)21(24(81-31)26(43)44)79-29-10(34-86(51,52)53)19(82-90(63,64)65)18(7(75-29)4-72-89(60,61)62)77-30-15(39)14(38)20(23(80-30)25(41)42)78-28-8(32-84(45,46)47)12(36)11(35)5(73-28)2-70-87(54,55)56;;;;;;;;;;/h5-24,27-40H,2-4H2,1H3,(H,41,42)(H,43,44)(H,45,46,47)(H,48,49,50)(H,51,52,53)(H,54,55,56)(H,57,58,59)(H,60,61,62)(H,63,64,65)(H,66,67,68);;;;;;;;;;/q;10*+1/p-10/t5-,6-,7-,8-,9-,10-,11-,12-,13-,14-,15-,16+,17-,18-,19-,20+,21+,22-,23+,24-,27+,28-,29-,30-,31-;;;;;;;;;;/m1........../s1

InChI key

XEKSTYNIJLDDAZ-JASSWCPGSA-D

Looking for similar products? Visit Product Comparison Guide

Application

Fondaparinux sodium has been used to test its neutralizing effect towards enterovirus D68-947 infection. It may be used in ultraviolet photodissociation (UVPD) measurements.

Biochem/physiol Actions

Fondaparinux sodium is an antithrombotic anticoagulant, a Factor Xa inhibitor.
Fondaparinux sodium is an antithrombotic anticoagulant, a Factor Xa inhibitor. Fondaparinux sodium is chemically related to low molecular weight heparins. Its pentasaccharide structure corresponds to the antithrombin III (ATIII) binding site of heparin. Fondaparinux sodium binding at this site potentiates the natural inhibitory effect of ATIII against factor Xa by a factor of approximately 300, which results in inhibition of thrombin generation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Yang et al.
Journal of clinical pharmacology, 60(12), 1671-1678 (2020-07-09)
Hepatotoxicity with low-molecular-weight heparin (LMWH) or fondaparinux is a relatively common adverse reaction. This study assessed the effects of LMWH and fondaparinux on liver function in patients with pulmonary embolism based on a retrospective cohort. As a result, a total
Dustin R Klein et al.
Analytical chemistry, 91(9), 6019-6026 (2019-04-02)
Structural characterization of sulfated glycosaminoglycans (GAGs) by mass spectrometry has long been a formidable analytical challenge owing to their high structural variability and the propensity for sulfate decomposition upon activation with low-energy ion activation methods. While derivatization and complexation workflows
Qing Guo et al.
Analytical and bioanalytical chemistry, 411(20), 5033-5045 (2019-06-04)
Glycosaminoglycans (GAGs) are carbohydrate polyionic polymers that participate in a host of critically important biological processes. A significant difficulty in the comprehensive structural characterization of GAGs is the determination of specific sulfation position isomers. We chose to circumvent sulfate lability
Emad Abdallah et al.
Obesity surgery, 30(10), 3735-3741 (2020-06-05)
Laparoscopic sleeve gastrectomy (LSG) is an effective bariatric procedure, yet can be followed by complications such as staple line leak and bleeding, vomiting, and gastroesophageal reflux disease (GERD). Various attempts have been described in the literature to improve the early
Li Fu et al.
Advanced drug delivery reviews, 97, 237-249 (2015-11-12)
Heparin and heparan sulfates are closely related linear anionic polysaccharides, called glycosaminoglycans, which exhibit a number of important biological and pharmacological activities. These polysaccharides, having complex structures and polydispersity, are biosynthesized in the Golgi of animal cells. While heparan sulfate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service