Skip to Content
Merck
All Photos(1)

Key Documents

SML1112

Sigma-Aldrich

Remodelin hydrobromide

≥98% (HPLC)

Synonym(s):

4-[2-(2-Cyclopentylidenehydrazinyl)-4-thiazolyl]-benzonitrile hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N4S · HBr
CAS Number:
Molecular Weight:
363.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

N#CC(C=C1)=CC=C1C2=CSC(NN=C3CCCC3)=N2.Br

InChI

1S/C15H14N4S.BrH/c16-9-11-5-7-12(8-6-11)14-10-20-15(17-14)19-18-13-3-1-2-4-13;/h5-8,10H,1-4H2,(H,17,19);1H

InChI key

XNWBCMSPDCSWSD-UHFFFAOYSA-N

Biochem/physiol Actions

Remodelin is an inhibitor of acetyl-transferase NAT10, a nucleolar N-acetyltransferase involved in stabilization of microtubules.
Remodelin is an inhibitor of acetyl-transferase NAT10, a nucleolar N-acetyltransferase involved in stabilization of microtubules. Remodelin was found to correct cell defects associated with progeria, improving nuclear shape and reducing the DNA damage believed to be associated with mutations in the gene for laminin A. Although the mechanism is still being studied, microtubule stability appears to be involved. Remodelin inhibition of NAT10 activity in laminopathic cells appears to result in reduced microtubule anchorage, normalizing the nuclear shape of laminopathic cells. Remodelin should be a useful tool to study how NAT10 affects nuclear architecture and its relationship to and possible treatments for laminopathies and aging and as well as its over-expression found in a variety of soft tissue sarcomas.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin Tsai et al.
Cell host & microbe, 28(2), 306-312 (2020-06-14)
Epitranscriptomic RNA modifications, including methylation of adenine and cytidine residues, are now recognized as key regulators of both cellular and viral mRNA function. Moreover, acetylation of the N4 position of cytidine (ac4C) was recently reported to increase the translation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service