Skip to Content
Merck
All Photos(1)

Key Documents

SML0953

Sigma-Aldrich

Cirsiliol

≥90% (HPLC)

Synonym(s):

2-(3,4-Dihydroxyphenyl)-5-hydroxy-6,7-dimethoxy-4H-1-benzopyran-4-one, 3′,4′,5-Trihydroxy-6,7-dimethoxy-flavone, 5,3′,4′-Trihydroxy-6,7-dimethoxyflavone, 6,7-Dimethoxy-5,3′,4′-trihydroxyflavone, 6-Hydroxyluteolin-6,7-dimethyl ether, 6-Methoxyluteolin 7-methyl ether, Crisiliol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C17H14O7/c1-22-14-7-13-15(16(21)17(14)23-2)11(20)6-12(24-13)8-3-4-9(18)10(19)5-8/h3-7,18-19,21H,1-2H3

InChI key

IMEYGBIXGJLUIS-UHFFFAOYSA-N

Biochem/physiol Actions

Cirsiliol is a flavonoid that displays anti-inflammatory, anti-oxidant, anti-proliferative, and anti-bacterial activities. Cirsiliol sensitizes non-small cell lung cancer cell lines (NSCLC) to radiation therapy. Cirsiliol inhibits epithelial-mesenchymal transition likely by induction of tumor-suppressive miR-34a, which suppresses Notch-1 expression NSCLC.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Horie et al.
Journal of medicinal chemistry, 29(11), 2256-2262 (1986-11-01)
Arachidonate 5-lipoxygenase plays a pivotal role in the biosynthesis of leukotrienes. Cirsiliol (3',4',5-trihydroxy-6,7-dimethoxyflavone), a selective inhibitor of the enzyme, was derivatized by introducing alkyl groups of various chain lengths at positions 5, 6, 7, and 8 of the A ring
Potent and selective 5-lipoxygenase inhibitors: cirsiliol and AA-861.
S W Kohno et al.
Advances in prostaglandin, thromboxane, and leukotriene research, 15, 217-219 (1985-01-01)
T Yoshimoto et al.
Biochemical and biophysical research communications, 116(2), 612-618 (1983-10-31)
Various flavonoids were found to be relatively selective inhibitors of arachidonate 5-lipoxygenase which initiates the biosynthesis of leukotrienes with the activity of slow reacting substance of anaphylaxis. Cirsiliol (3',4',5-trihydroxy-6,7-dimethoxyflavone) was most potent, and the enzyme partially purified from rat basophilic
Mariel Marder et al.
Current topics in medicinal chemistry, 2(8), 853-867 (2002-08-13)
This review describes the new research developments that have established the CNS-activity of some natural flavonoids. The properties of flavone, chrysin, apigenin and cirsiliol are described and a survey of the occurrence of ligands for the benzodiazepine binding site in
S Yamamoto et al.
The Journal of allergy and clinical immunology, 74(3 Pt 2), 349-352 (1984-09-01)
The 5-lipoxygenases of guinea pig peritoneal polymorphonuclear leukocytes and of rat basophilic leukemia cells have been solubilized, purified partially by affinity chromatography, and shown to convert arachidonic acid principally to 5-hydroperoxy-6,8,11,14- eicosatetraenoic acid. The activity of both 5-lipoxygenases is calcium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service