Skip to Content
Merck
All Photos(1)

Key Documents

SML0911

Sigma-Aldrich

TAK-779

≥98% (HPLC)

Synonym(s):

N,N-Dimethyl-N-[4-[[[2-(4-methylphenyl)-6,7-dihydro-5H-benzohepten-8-yl]carbonyl]amino]benzyl]tetrahydro-2H-pyran-4-aminium chloride, TAK-799, Takeda 779

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H39ClN2O2
CAS Number:
Molecular Weight:
531.13
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Cl.[N+](C5CCOCC5)(Cc1ccc(cc1)\N=C(/[O-])\C2=Cc3c(ccc(c3)c4ccc(cc4)C)CCC2)(C)C

InChI

1S/C33H38N2O2.ClH/c1-24-7-11-27(12-8-24)28-14-13-26-5-4-6-29(22-30(26)21-28)33(36)34-31-15-9-25(10-16-31)23-35(2,3)32-17-19-37-20-18-32;/h7-16,21-22,32H,4-6,17-20,23H2,1-3H3;1H

InChI key

VDALIBWXVQVFGZ-UHFFFAOYSA-N

Application

TAK-779 has been used to inhibit CC chemokine ligand 5 (CCL5)– C-C chemokine receptor type 5 (CCR5) interaction in vitro in natural killer (NK) cytotoxicity assay. It has also been used as a blocker of CCR5 to study its effects and to evaluate the opening of the Panx-1 channels on peripheral blood mononuclear cells (PBMCs) isolated from human immunodeficiency virus (HIV)-infected individuals.

Biochem/physiol Actions

TAK-779 inhibits human immunodeficiency virus (HIV) entrance by blocking the C-C chemokine receptor type 5 (CCR5). It also impairs the formation of atherosclerotic lesions by preventing T-helper 1 cells from entering the plaque. TAK-779 possesses anti-immunogenic properties. In an arthritis model, it could prevent the inflow of CCR5- and chemokine (C-X-C motif) receptor 3 (CXCR3)-positive T cells into inflamed joints. In mice, TTAK-779 protects the brain from focal cerebral ischemia.
TAK-779 is a potent, dual antagonist at chemokine receptors C-C chemokine receptor type 2 (CCR2) and C-C chemokine receptor type 5 (CCR5) with IC50 = 1.4 nM at CCR5 and 2.3 nM at CCR2. Antagonists of both CCR2 and CCR5 such as TAK-779 have been investigated for treatment of viruses, rheumatoid arthritis, multiple sclerosis, and cancer. CCR5 is particularly targeted for anti-HIV therapy, since HIV entry into cells requires chemokine coreceptors CCR5 and C-X-C motif chemokine receptor 4 (CXCR4).
TAK-779 is a potent, dual antagonist at chemokine receptors CCR2 and CCR5.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hengjie Xu et al.
Transplantation, 86(12), 1810-1817 (2008-12-24)
The effect of blocking lymphocyte chemokine receptors with TAK-779 was investigated using a rat intestinal transplantation model. Dark Agouti rat small intestines were heterotopically transplanted into Lewis rats. The recipients were treated with TAK-779 (10 mg/kg per day) by subcutaneous
Shinya Takami et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 22(7), 780-784 (2002-07-27)
The effect of a nonpeptide CC chemokine receptor antagonist, TAK-779, on ischemic brain injury resulting from 1-hour middle cerebral artery occlusion followed by 48-hour reperfusion was examined in ddY mice. On intracerebroventricular injection of vehicle or TAK-779, infarct volume in
Fernando Arenzana-Seisdedos
Enfermedades infecciosas y microbiologia clinica, 26 Suppl 11, 5-11 (2009-03-14)
Viral entry is an early stage and specific of the infection in which different viral and cellular targets are accessible to therapeutic treatment. CXCR4 and CCR5 act in this process as coreceptor molecules of HIV for its entry into the
Theresa L Chang et al.
Journal of acquired immune deficiency syndromes (1999), 53(3), 292-302 (2010-01-13)
Macrophages are major HIV target cells. They support both productive and latent HIV-1 infection. Susceptibility of primary macrophages to HIV depends on the anatomical location and activation state of the cells. We demonstrate that peritoneal macrophages (PMs) are abundant in
Qinxue Hu et al.
The Journal of general virology, 91(Pt 12), 2965-2973 (2010-09-03)
The envelope glycoprotein (Env) of human immunodeficiency virus is key to viral entry of susceptible target cells and is therefore a major target for the design of vaccines and antiviral drugs. C-C chemokine receptor type 5 (CCR5)-using (R5) Env is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service