Skip to Content
Merck
All Photos(1)

Documents

SML0789

Sigma-Aldrich

GI254023X

≥98% (HPLC), powder, ADAM10 metalloproteinase inhibitor

Synonym(s):

(2R)-N-[(1S)-2,2-Dimethyl-1-[(methylamino)carbonyl]-propyl]-2-[(1S)-1-[formyl(hydroxy)amino]ethyl]-5-phenylpentanamide, GI4023

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H33N3O4
CAS Number:
Molecular Weight:
391.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

GI254023X, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

ON(C=O)[C@@H](C)[C@@H](CCCC1=CC=CC=C1)C(N[C@@H](C(C)(C)C)C(NC)=O)=O

InChI

1S/C21H33N3O4/c1-15(24(28)14-25)17(13-9-12-16-10-7-6-8-11-16)19(26)23-18(20(27)22-5)21(2,3)4/h6-8,10-11,14-15,17-18,28H,9,12-13H2,1-5H3,(H,22,27)(H,23,26)/t15-,17+,18+/m0/s1

InChI key

GHVMTHKJUAOZJP-CGTJXYLNSA-N

Application

GI254023X has been used to inhibit ADAM10 (ADAM metallopeptidase domain 10).

Biochem/physiol Actions

GI254023X blocks ADAM10 (ADAM metallopeptidase domain 10) activity and decreases human leukocyte antigen (HLA)-mediated cytotoxicity and cleavage of extracellular E-cadherin in epithelial and endothelial cells.
GI254023X is a potent and selective ADAM10 metalloproteinase inhibitor with 100-fold selectivity for the α-secretase ADAM10 over ADAM17 (TACE). In a study using recombinant TACE and ADAM10 ectodomain, the IC50 for GI254023X was 5.3 nM for ADAM10 vs 541 nM for TACE.
GI254023X is a potent and selective ADAM10 metalloproteinase inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ADAM10 cell surface expression but not activity is critical for Staphylococcus aureus α-hemolysin-mediated activation of the NLRP3 inflammasome in human monocytes.
Ezekwe E A D, et al.
Toxins, 8(4), 95-95 (2016)
Kazuhiro Aoki et al.
Developmental cell, 43(3), 305-317 (2017-11-08)
The biophysical framework of collective cell migration has been extensively investigated in recent years; however, it remains elusive how chemical inputs from neighboring cells are integrated to coordinate the collective movement. Here, we provide evidence that propagation waves of extracellular
The Mouse-specific Splice Variant mRAGE_v4 Encodes a Membrane-bound RAGE that is Resistant to Shedding and does not Contribute to the Production of Soluble RAGE.
Di Maggio S, et al.
PLoS ONE, 11(9), e0153832-e0153832 (2016)
Joshua A Kulas et al.
American journal of physiology. Endocrinology and metabolism, 316(1), E106-E120 (2018-11-14)
The amyloid precursor protein (APP) is a type I transmembrane glycoprotein widely studied for its role as the source of β-amyloid peptide, accumulation of which is causal in at least some cases of Alzheimer's disease (AD). APP is expressed ubiquitously
Laura S Christian et al.
Cell reports, 35(6), 109118-109118 (2021-05-13)
As a critical machinery for rapid pathogen removal, resident memory T cells (TRMs) are locally generated after the initial encounter. However, their development accompanying tumorigenesis remains elusive. Using a murine breast cancer model, we show that TRMs develop in the tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service