Skip to Content
Merck
All Photos(1)

Key Documents

579050

Sigma-Aldrich

TAPI-0

≥96% (HPLC), solid, collagenase, gelatinase, and TACE inhibitor, Calbiochem®

Synonym(s):

TAPI-0, N-(R)-[2-(Hydroxyaminocarbonyl)methyl]-4-methylpentanoyl-L-naphthylalanyl-L-alanine Amide, TNF-α Protease Inhibitor-0

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32N4O5
CAS Number:
Molecular Weight:
456.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

TAPI-0, TAPI-0, CAS 143457-40-3, is a hydroxamate-based inhibitor of collagenase, gelatinase, and TACE (TNF-α converting enzyme/ADAM17; IC₅₀ = 100 nM).

Quality Level

Assay

≥96% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

50% acetic acid: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N(O)C(=O)CC(CC(C)C)C(=O)N[C@@H](Cc1cc2c(cc1)cccc2)C(=O)N[C@@H](C)C(=O)N

InChI

1S/C24H32N4O5/c1-14(2)10-19(13-21(29)28-33)23(31)27-20(24(32)26-15(3)22(25)30)12-16-8-9-17-6-4-5-7-18(17)11-16/h4-9,11,14-15,19-20,33H,10,12-13H2,1-3H3,(H2,25,30)(H,26,32)(H,27,31)(H,28,29)/t15-,19?,20-/m0/s1

InChI key

CRCPLBFLOSEABN-VBSNWNEZSA-N

General description

A hydroxamate-based inhibitor of collagenase, gelatinase, and TACE (TNF-α converting enzyme/ADAM17; IC50 = 100 nM). Blocks spontaneous and PMA-induced release of TNF-α in transfected COS 7 cells.
A hydroxymate-based inhibitor of collagenase, gelatinase, and TACE [(TNF-α convertase; ADAM17 (IC50 = 100 nM)]. Blocks spontaneous and PMA-induced release of TNF-α from transfected COS-7 cells.

Biochem/physiol Actions

Cell permeable: no
Primary Target
TNF-α converting enzyme/ADAM17
Product does not compete with ATP.
Reversible: no
Target IC50: 100 nM against TACE (TNF-α converting enzyme/ADAM17)

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Hooper, N.M., et al. 1997. Biochem. J.321, 265.
Mullberg, J., et al. 1995. J. Immunol.155, 5198.
Mohler, K.M., et al. 1994. Nature370, 218.

Legal Information

Manufactured exclusively by Peptides International, Louisville, Kentucky, USA.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sandra C Côté et al.
European journal of immunology, 50(10), 1537-1549 (2020-05-12)
The IL-7 receptor specific α chain, CD127, can be expressed both as a membrane-associated (mCD127) and a soluble form (sCD127), however, the mechanisms involved in their regulation remain to be defined. We first demonstrated in primary human CD8+ T cells
Youngkyung Cho et al.
BMC biology, 18(1), 60-60 (2020-06-05)
Epithin/PRSS14, a type II transmembrane serine protease, is an emerging target of cancer therapy because of its critical roles in tumor progression and metastasis. In many circumstances, the protease, through its ectodomain shedding, exists as a soluble form and performs
Alice Liu et al.
PloS one, 15(7), e0236175-e0236175 (2020-07-23)
Adenoviruses cause upper respiratory infections, conjunctivitis, keratitis, and gastrointestinal illness. These can be fatal in immunocompromised individuals. Adenoviruses have also been engineered into viral vectors to deliver therapeutic genes or induce immunity as vaccine carriers. The success of ocular gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service