Skip to Content
Merck
All Photos(1)

Key Documents

SML0627

Sigma-Aldrich

ML-SA1

≥95% (HPLC), powder, TRPML channel agonist

Synonym(s):

1-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetyl]-1,2,3,4-tetrahydro-2,2,4-trimethyl-quinoline, 2-(2-Oxo-2-(2,2,4-trimethyl-3,4-dihydroquinolin-1(2H)-yl)ethyl)isoindoline-1,3-dione, Mucolipin Synthetic Agonist 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N2O3
CAS Number:
Molecular Weight:
362.42
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

ML-SA1, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

N3(C(=O)c4c(cccc4)C3=O)CC(=O)N1C(CC(c2c1cccc2)C)(C)C

InChI

1S/C22H22N2O3/c1-14-12-22(2,3)24(18-11-7-6-8-15(14)18)19(25)13-23-20(26)16-9-4-5-10-17(16)21(23)27/h4-11,14H,12-13H2,1-3H3

InChI key

KDDHBJICVBONAX-UHFFFAOYSA-N

Application

ML-SA1 has been used as a transient receptor potential cation channel mucolipin 1 (TRPML1/ML1) agonist.

Biochem/physiol Actions

ML-SA1 is a potent and selective cell permeable agonist of lysosomal mucolipin transient receptor potential channels (TRPML) 1, 2, 3 that significantly increases [Ca2+]cyt in HEK293 cells stably- or transiently-expressing mutant TRPML1 channels ML1-4A.
ML-SA1 is a potent and selective cell permeable agonist of lysosomal mucolipin transient receptor potential channels (TRPML) 1, 2, 3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gastric acid secretion from parietal cells is mediated by a Ca2+ efflux channel in the tubulovesicle.
Sahoo N, et al.
Developmental Cell, 41(3), 262-273 (2017)
Dengqin Zhong et al.
Nature communications, 14(1), 3997-3997 (2023-07-07)
Uranium (U) is a well-known nephrotoxicant which forms precipitates in the lysosomes of renal proximal tubular epithelial cells (PTECs) after U-exposure at a cytotoxic dose. However, the roles of lysosomes in U decorporation and detoxification remain to be elucidated. Mucolipin
Hongyan Yu et al.
British journal of cancer, 125(10), 1420-1431 (2021-09-23)
Prostate cancer (Pca) is the most common cancer type among males worldwide. Dysregulation of Ca2+ signaling plays important roles during Pca progression. However, there is lack of information about the role of endolysosomal Ca2+ -permeable channels in Pca progression. The
Simona Ida Scorza et al.
International journal of molecular sciences, 24(2) (2023-01-22)
Lysosomes are acidic Ca2+ storage organelles that actively generate local Ca2+ signaling events to regulate a plethora of cell functions. Here, we characterized lysosomal Ca2+ signals in mouse renal collecting duct (CD) cells and we assessed their putative role in
Dale P Corkery et al.
EMBO reports, 24(9), e56841-e56841 (2023-06-29)
Lysosomal membrane damage represents a threat to cell viability. As such, cells have evolved sophisticated mechanisms to maintain lysosomal integrity. Small membrane lesions are detected and repaired by the endosomal sorting complex required for transport (ESCRT) machinery while more extensively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service