Skip to Content
Merck
All Photos(1)

Key Documents

SML0406

Sigma-Aldrich

Roquefortine C

≥98% (HPLC), from Penicillium roqueforti

Synonym(s):

2H-Pyrazino[1′,2′:1,5]pyrrolo[2,3-b]indole-1,4(3H,5aH)-dione, 10b-(1,1-dimethyl-2-propen-1-yl)-6,10b,11,11a-tetrahydro-3-(1H-imidazol-5-ylmethylene)-,(3E,5aS,10bR,11aS)-, Roquefortine from Penicillium roqueforti

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N5O2
CAS Number:
Molecular Weight:
389.45
UNSPSC Code:
12161501
NACRES:
NA.77

biological source

Penicillium roqueforti

Quality Level

Assay

≥98% (HPLC)

solubility

chloroform: 1 mg/mL
ethyl acetate: 1 mg/mL
DMSO: 10 mg/mL
methanol: 10 mg/mL

storage temp.

−20°C

SMILES string

N21C3Nc4c(cccc4)C3(CC2C(=O)N\C(=C/c5nc[nH]c5)\C1=O)C(C)(C)C=C

InChI

1S/C22H23N5O2/c1-4-21(2,3)22-10-17-18(28)25-16(9-13-11-23-12-24-13)19(29)27(17)20(22)26-15-8-6-5-7-14(15)22/h4-9,11-12,17,20,26H,1,10H2,2-3H3,(H,23,24)(H,25,28)/b16-9-

InChI key

SPWSUFUPTSJWNG-SXGWCWSVSA-N

Application

Roquefortine C has been used as a standard for the quantification of roquefortine C by high-performance liquid chromatography (HPLC). It has also been used as a standard for the quantification of roquefortine C by liquid chromatography-mass spectrometry (LC−MS/MS).

Biochem/physiol Actions

Roquefortine C is a paralytic neurotoxin of a dioxopiperazine structure produced by a diverse range of fungi, most notably Penicillium species.
Roquefortine C is a paralytic neurotoxin of a dioxopiperazine structure produced by a diverse range of fungi, most notably Penicillium species. It has been found in blue cheese and in many other food products due to natural occurrence and contamination. Roquefortine C was found to be active on a wide range of organisms. It inhibits the growth of Gram-positive bacteria, and cockerels treated with roquefortine lost their righting reflex and died within 8-12 hours. Mice injected with roquefortine C experienced neurotoxic properties. Roquefortine C was also reported to inhibit cytochrome P450 as well as tubulin polymerization.

Preparation Note

Soluble in methanol (10 mg/mL), DMSO (10 mg/mL), ethyl acetate (1 mg/mL) and chloroform (1 mg/mL). DMSO solution at 10 mg/mL is stable for 3 months at −20 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A natural short pathway synthesizes roquefortine C but not meleagrin in three different Penicillium roqueforti strains
Kosalkova K, et al.
Applied Microbiology and Biotechnology, 99(18), 7601-7612 (2015)
David J Richard et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(33), 11971-11976 (2004-05-14)
The syntheses of isoroquefortine C and a related heterocycle were achieved by implementation of both intra- and intermolecular vinyl amidation reactions. These accomplishments represent a significant advance in the use of these strategies in the generation of complex molecules.
Katarina Kosalková et al.
Biochimie, 91(2), 214-225 (2008-10-28)
The biosynthesis of the beta-lactam antibiotic penicillin is an excellent model for the study of secondary metabolites produced by filamentous fungi due to the good background knowledge on the biochemistry and molecular genetics of the beta-lactam producing microorganisms. The three
Ramón O García-Rico et al.
International microbiology : the official journal of the Spanish Society for Microbiology, 12(2), 123-129 (2009-09-29)
Heterotrimeric G protein signaling regulates many processes in fungi, such as development, pathogenicity, and secondary metabolite biosynthesis. For example, the Galpha subunit Pga1 from Penicillium chrysogenum regulates conidiation and secondary metabolite production in this fungus. The dominant activating allele, pga1G42R
C Aninat et al.
Food additives and contaminants, 22(4), 361-368 (2005-07-16)
Man is permanently exposed to exogenous substances, either natural ones (e.g. mycotoxins, plant extracts) or man-made compounds such as pesticides or drugs. In some cases, such foreign compounds can exert either therapeutic (drugs) or toxic effects, or both. In particular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service