Skip to Content
Merck
All Photos(1)

Key Documents

SML0364

Sigma-Aldrich

RAD51 Inhibitor B02

≥98% (HPLC)

Synonym(s):

3-(Phenylmethyl)-2-[(1E)-2-(3-pyridinyl)ethenyl]-4(3H)-quinazolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17N3O
CAS Number:
Molecular Weight:
339.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to dark brown

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

O=C1N(CC2=CC=CC=C2)C(/C=C/C3=CN=CC=C3)=NC4=C1C=CC=C4

InChI

1S/C22H17N3O/c26-22-19-10-4-5-11-20(19)24-21(13-12-17-9-6-14-23-15-17)25(22)16-18-7-2-1-3-8-18/h1-15H,16H2/b13-12+

InChI key

GEKDQXSPTHHANP-OUKQBFOZSA-N

Application

RAD51 Inhibitor B02 has been used:
  • to test its effect on the polar body extrusion (PBE) rate in porcine oocytes
  • for RAD51 inhibition in porcine embryos
  • as RAD51 inhibitor and to test its effect on targeted nucleotide substitution (TNS) in induced pluripotent stem cells (iPSCs)

Biochem/physiol Actions

B02 (3-(Phenylmethyl)-2-[(1E)-2-(3-pyridinyl)ethenyl]-4(3H)-quinazolinone), a pyridinylvinyl-quinazolinone compound is cell-permeable. B02 inhibits human RAD51 recombinase and subsequent nucleofilaments formation. It halts homologous recombination (HR) repair events in cancer cells. B02 favors apoptosis in multiple myeloma and is crucial for sensitizing them to doxorubicin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A small-molecule inhibitor of RAD51 reduces homologous recombination and sensitizes multiple myeloma cells to doxorubicin
Alagpulinsa DA, et al.
Frontiers in Oncology, 4, 289-289 (2014)
Ivana Murfuni et al.
PLoS genetics, 9(10), e1003910-e1003910 (2013-11-10)
In checkpoint-deficient cells, DNA double-strand breaks (DSBs) are produced during replication by the structure-specific endonuclease MUS81. The mechanism underlying MUS81-dependent cleavage, and the effect on chromosome integrity and viability of checkpoint deficient cells is only partly understood, especially in human
Zhe-Long Jin et al.
Reproduction (Cambridge, England), 157(3), 223-234 (2019-03-01)
Homologous recombination (HR) plays a critical role in facilitating replication fork progression when the polymerase complex encounters a blocking DNA lesion, and it also serves as the primary mechanism for error-free DNA repair of double-stranded breaks. DNA repair protein RAD51
Arthur Aubry et al.
Oncogene, 39(31), 5338-5357 (2020-06-24)
Local intravitreal or intra-arterial chemotherapy has improved therapeutic success for the pediatric cancer retinoblastoma (RB), but toxicity remains a major caveat. RB initiates primarily with RB1 loss or, rarely, MYCN amplification, but the critical downstream networks are incompletely understood. We
Inhibition of DNA repair protein RAD51 affects porcine preimplantation embryo development
Jin ZL, et al.
Reproduction (Cambridge, England), 157(3) (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service