Skip to Content
Merck
All Photos(1)

Key Documents

SML0355

Sigma-Aldrich

BCTC

≥98% (HPLC)

Synonym(s):

4-(3-Chloro-2-pyridinyl)-N-[4-(1,1-dimethylethyl)phenyl]-1-piperazinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25ClN4O
CAS Number:
Molecular Weight:
372.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

CC(C)(C)c1ccc(NC(=O)N2CCN(CC2)c3ncccc3Cl)cc1

InChI

1S/C20H25ClN4O/c1-20(2,3)15-6-8-16(9-7-15)23-19(26)25-13-11-24(12-14-25)18-17(21)5-4-10-22-18/h4-10H,11-14H2,1-3H3,(H,23,26)

InChI key

ROGUAPYLUCHQGK-UHFFFAOYSA-N

Application

BCTC or (4-Tertbutylphenyl)-4-(3-chloropyridin-2-Yl) tetrahydropyrazine-1(2H)-carbox-amide has been used as transient receptor potential melastatin 8 (TRPM8) antagonist to study its effects on proliferation and cell viability.

Biochem/physiol Actions

BCTC is a potent antagonist of the vanilloid receptor TRPV1. BCTC blocks capsaicin-induced contractions in a variety of smooth muscle tissues, including guinea pig ileum, bladder and trachea with no effects on capsaicin-sensitive, sensory neuron-mediated positive inotropy. The compound penetrates the CNS and has strong analgesic properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haipei Liu et al.
Inflammation, 41(4), 1266-1275 (2018-04-10)
Bronchial asthma is a chronic inflammatory airway disease that can be aggravated by cold air. However, its mechanism remains largely unknown. As a thermo-sensing cation channel, transient receptor potential melastatin 8 (TRPM8) can be activated by cold stimuli (8-22 °C) and
Andrea Fiebig et al.
Journal of translational medicine, 21(1), 908-908 (2023-12-13)
Chronic itch (chronic pruritus) is a major therapeutic challenge that remains poorly understood despite the extensive recent analysis of human pruriceptors. It is unclear how the peripheral nervous system differentiates the signaling of non-histaminergic itch and pain. Here we used
Gang Wang et al.
OncoTargets and therapy, 13, 8825-8835 (2020-09-19)
Based on accumulating evidence, transient receptor potential (TRP) ion channels may play important roles in the occurrence and the progression of cancer. TRP melastatin 8 (TRPM8), a member of the TRP family, functions as a Ca2+-permeable channel and regulates various
Corinna Martin et al.
Frontiers in molecular neuroscience, 11, 7-7 (2018-02-08)
Oxidized phospholipids (OxPL) like oxidized 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphocholine (OxPAPC) were recently identified as novel proalgesic targets in acute and chronic inflammatory pain. These endogenous chemical irritants are generated in inflamed tissue and mediate their pain-inducing function by activating the transient receptor potential
Yuki Nishizawa et al.
Journal of neuroscience research, 96(5), 901-910 (2017-12-29)
T-type Ca2+ channels and TRPA1 are expressed in sensory neurons and both are associated with pain transmission, but their functional interaction is unclear. Here we demonstrate that pharmacological evidence of the functional relation between T-type Ca2+ channels and TRPA1 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service