Skip to Content
Merck
All Photos(2)

Documents

P21005

Sigma-Aldrich

4-Phenylbutyric acid

99%

Synonym(s):

γ-Phenylbutanoic acid, ψ-Phenylbutanoic acid, 4-Phenyl-n-butyric acid, 4-Phenylbutanoic acid, Benzenebutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)3COOH
CAS Number:
Molecular Weight:
164.20
Beilstein:
638180
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

bp

165 °C/10 mmHg (lit.)

mp

49-51 °C (lit.)

SMILES string

OC(=O)CCCc1ccccc1

InChI

1S/C10H12O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2,(H,11,12)

InChI key

OBKXEAXTFZPCHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Phenylbutyric acid (4-PBA) is used in the treatment of urea cycle disorders.

Application

4-PBA has been used as a selective inhibitor of endoplasmic reticulum stress (ERS).
It can also be used as a reactant in the synthesis of:
  • 1-Tetralone using Lewis acid catalyst.
  • 4,N-diphenylbutyramide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Induction of apoptosis coupled to endoplasmic reticulum stress through regulation of CHOP and JNK in bone marrow mesenchymal stem cells from patients with systemic lupus erythematosus
Guo G, et al.
Journal of immunology research (2015)
Min Han Kim et al.
Clinical and experimental otorhinolaryngology, 12(2), 181-189 (2018-10-20)
Endoplasmic reticulum (ER) stress is known to be associated with inflammatory airway diseases, and three major transmembrane receptors: double-stranded RNA-activated protein kinase-like ER kinase, inositol requiring enzyme 1, and activating transcription factor 6 (ATF6) play important roles in ER stress-related
Synthesis of 1-tetralones by intramolecular Friedel?Crafts reaction of 4-arylbutyric acids using Lewis acid catalysts
Cui DM, et al.
Tetrahedron Letters, 44(21), 4007-4010 (2003)
Clean, reusable and low cost heterogeneous catalyst for amide synthesis
Comerford JW, et al.
Chemical Communications (Cambridge, England), 18(21), 2562-2564 (2009)
The therapeutic effects of 4-phenylbutyric acid in maintaining proteostasis.
Kolb PS, et al.
The International Journal of Biochemistry & Cell Biology, 61, 45-52 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service