
Tolcapone

Synonym(s):
3,4-Dihydroxy-4′-methyl-5-nitrobenzophenone, Ro 40-7592
Empirical Formula (Hill Notation):
C14H11NO5
CAS Number:
Molecular Weight:
273.24
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing